missing translation for 'onlineSavingsMsg'
Learn More

4'-(Trifluoromethoxy)acetophenone 98.0+%, TCI America™

Supplier:  TCI America T19405G

Catalog No. T19405G

Only null left
Explore available promotions

Description

Description

Specifications

Specifications

4′-(Trifluoromethoxy)acetophenone
Yellow
MFCD00042404
4'-trifluoromethoxy acetophenone, 1-4-trifluoromethoxy phenyl ethanone, 4-trifluoromethoxy acetophenone, 1-4-trifluoromethoxy phenyl ethan-1-one, 4'-trifluoromethoxyacetophenone, 1-4-trifluoromethoxy-phenyl-ethanone, ethanone, 1-4-trifluoromethoxy phenyl, 4-trifluoromethoxyacetophenone
CC(=O)C1=CC=C(OC(F)(F)F)C=C1
204.15
204.15
Liquid
85013-98-5
C9H7F3O2
5 g
MOEXTBIPPMLEFX-UHFFFAOYSA-N
1-[4-(trifluoromethoxy)phenyl]ethan-1-one
522821
≥98.0% (GC)

Chemical Identifiers

85013-98-5
204.15
MOEXTBIPPMLEFX-UHFFFAOYSA-N
522821
CC(=O)C1=CC=C(OC(F)(F)F)C=C1
C9H7F3O2
MFCD00042404
4'-trifluoromethoxy acetophenone, 1-4-trifluoromethoxy phenyl ethanone, 4-trifluoromethoxy acetophenone, 1-4-trifluoromethoxy phenyl ethan-1-one, 4'-trifluoromethoxyacetophenone, 1-4-trifluoromethoxy-phenyl-ethanone, ethanone, 1-4-trifluoromethoxy phenyl, 4-trifluoromethoxyacetophenone
1-[4-(trifluoromethoxy)phenyl]ethan-1-one
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
Documents

Documents

Promotions

Promotions