missing translation for 'onlineSavingsMsg'
Learn More

4-(Trifluoroacetyl)toluene, 97%

Catalog No. AAL1242403
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1242403 1 g
AAL1242406 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1242403 Supplier Thermo Scientific Chemicals Supplier No. L1242403

CAS: 394-59-2 | C9H7F3O | 188.149 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 394-59-2
Molecular Formula C9H7F3O
Molecular Weight (g/mol) 188.149
MDL Number MFCD00052338
InChI Key DYILUJUELMWXAL-UHFFFAOYSA-N
Synonym 4-trifluoroacetyl toluene, 2,2,2-trifluoro-1-p-tolyl ethanone, 4'-methyl-2,2,2-trifluoroacetophenone, 2,2,2-trifluoro-1-p-tolylethanone, 2,2,2-trifluoro-1-4-methylphenyl ethanone, 2,2,2-trifluoro-4'-methylacetophenone, 2,2,2-trifluoro-1-4-methylphenyl ethan-1-one, 2,2,2-trifluoro-1-p-tolyl-ethanone, 4-trifluoroacetyltoluene, acmc-209j5m
PubChem CID 136237
IUPAC Name 2,2,2-trifluoro-1-(4-methylphenyl)ethanone
SMILES CC1=CC=C(C=C1)C(=O)C(F)(F)F

Specifications

Boiling Point 70°C to 72°C (15 mmHg)
Refractive Index 1.47
Quantity 1 g
Beilstein 1870406
Formula Weight 188.15
Percent Purity 97%
Chemical Name or Material 4-(Trifluoroacetyl)toluene
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only