missing translation for 'onlineSavingsMsg'
Learn More

4-(Trifluoroacetyl)benzoic acid, 97+%

CAS: 58808-59-6 | C9H5F3O3 | 218.131 g/mol

$86.33 - $199.48

Chemical Identifiers

CAS 58808-59-6
Molecular Formula C9H5F3O3
Molecular Weight (g/mol) 218.131
MDL Number MFCD00052340
InChI Key WLTZCRCZDLLXQP-UHFFFAOYSA-N
Synonym 4-trifluoroacetyl benzoic acid, 4-2,2,2-trifluoroacetyl benzoic acid, benzoic acid, 4-trifluoroacetyl, 4-2,2,2-trifluoro-acetyl-benzoic acid, benzoic acid, 4-trifluoroacetyl-9ci, 4-carboxy-a,a,a-trifluoroacetophenone, pubchem13997, acmc-1atst, 4-trifluoroacetylbenzoic acid, labotest-bb lt00454127
PubChem CID 143579
IUPAC Name 4-(2,2,2-trifluoroacetyl)benzoic acid
SMILES C1=CC(=CC=C1C(=O)C(F)(F)F)C(=O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL10242MD
View Documents
Thermo Scientific Chemicals
L10242MD
250 mg
Each for $86.33
Only null left
 
AAL1024203
View Documents
Thermo Scientific Chemicals
L1024203
1 g
Each for $199.48
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

58808-59-6
218.131
WLTZCRCZDLLXQP-UHFFFAOYSA-N
143579
C1=CC(=CC=C1C(=O)C(F)(F)F)C(=O)O
C9H5F3O3
MFCD00052340
4-trifluoroacetyl benzoic acid, 4-2,2,2-trifluoroacetyl benzoic acid, benzoic acid, 4-trifluoroacetyl, 4-2,2,2-trifluoro-acetyl-benzoic acid, benzoic acid, 4-trifluoroacetyl-9ci, 4-carboxy-a,a,a-trifluoroacetophenone, pubchem13997, acmc-1atst, 4-trifluoroacetylbenzoic acid, labotest-bb lt00454127
4-(2,2,2-trifluoroacetyl)benzoic acid

Specifications

58808-59-6
C9H5F3O3
250 mg
Hygroscopic
WLTZCRCZDLLXQP-UHFFFAOYSA-N
4-(2,2,2-trifluoroacetyl)benzoic acid
143579
≥97%
177°C to 179°C
MFCD00052340
2728778
4-trifluoroacetyl benzoic acid, 4-2,2,2-trifluoroacetyl benzoic acid, benzoic acid, 4-trifluoroacetyl, 4-2,2,2-trifluoro-acetyl-benzoic acid, benzoic acid, 4-trifluoroacetyl-9ci, 4-carboxy-a,a,a-trifluoroacetophenone, pubchem13997, acmc-1atst, 4-trifluoroacetylbenzoic acid, labotest-bb lt00454127
C1=CC(=CC=C1C(=O)C(F)(F)F)C(=O)O
218.131
218.13
4-(Trifluoroacetyl)benzoic acid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures; Store under Nitrogen

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only