missing translation for 'onlineSavingsMsg'
Learn More

4-Trifluoroacetyl-3-methyl-1-phenyl-5-pyrazolone 96.0+%, TCI America™

Catalog No. T12865G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T12865G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T12865G Supplier TCI America Supplier No. T12865G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1691-93-6
Molecular Formula C12H9F3N2O2
Molecular Weight (g/mol) 270.211
MDL Number MFCD00143378
InChI Key XJJDGTPFEFAAMV-UHFFFAOYSA-N
Synonym 3-Methyl-1-phenyl-4-trifluoroacetyl-5-pyrazolone
PubChem CID 781778
IUPAC Name 5-methyl-2-phenyl-4-(2,2,2-trifluoroacetyl)-1H-pyrazol-3-one
SMILES CC1=C(C(=O)N(N1)C2=CC=CC=C2)C(=O)C(F)(F)F

Specifications

Melting Point 147°C
Quantity 5 g
Formula Weight 270.21
Percent Purity ≥96.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Trifluoroacetyl-3-methyl-1-phenyl-5-pyrazolone