missing translation for 'onlineSavingsMsg'
Learn More

4-(Tri-n-butylstannyl)pyridine, 96%

Catalog No. p-7033989
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5143503 1 g
AAH5143506 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog No. AAH5143503 Supplier Thermo Scientific Chemicals Supplier No. H5143503
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.

CAS: 124252-41-1 | C17H31NSn | 368.15 g/mol

4-(Tri-n-butylstannyl)pyridine is used as organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-(Tri-n-butylstannyl)pyridine is used as organic chemical synthesis intermediate.

Solubility
Not miscible in water. Soluble in ether, hexane and tetrahydrofuran.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 124252-41-1
Molecular Formula C17H31NSn
Molecular Weight (g/mol) 368.15
MDL Number MFCD01319071
InChI Key UNEPXPMBVGDXGH-UHFFFAOYSA-N
PubChem CID 2737318
IUPAC Name tributyl(pyridin-4-yl)stannane
SMILES CCCC[Sn](CCCC)(CCCC)C1=CC=NC=C1

Specifications

Quantity 1 g
UN Number UN2788
Synonym 4-tributylstannyl pyridine, pyridine,4-tributylstannyl, 4-tributylstannylpyridine, 4-tributylstannyl-pyridine, 4-tri-n-butylstannyl pyridine, pyridin-4-yl tributylstannane, pyridine, 4-tributylstannyl, 4-pyridyltributyltin, 4-pyridyl tributyltin, 4-tributyistannylpyridine
Solubility Information Not miscible in water. Soluble in ether,hexane and tetrahydrofuran.
Formula Weight 368.15
Percent Purity 96%
Chemical Name or Material 4-(Tri-n-butylstannyl)pyridine
Hazard Category H301-H312-H315-H319-H372
Hazard Statement GHS H Statement
H301-H372-H312-H315-H319
Toxic if swallowed.
Causes damage to organs through prolonged or repeated exposure.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P260-P264b-P270-P280-P301+P310-P302+P352-P305+P351+P338-P312-P314-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ORGANOTIN COMPOUNDS, LIQUID, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only