missing translation for 'onlineSavingsMsg'
Learn More

4-(trans-4-Ethylcyclohexyl)cyclohexanone 98.0+%, TCI America™

Catalog No. E11975G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E11975G 5 g
E119725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E11975G Supplier TCI America Supplier No. E11975G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 150763-46-5
Molecular Formula C14H24O
Molecular Weight (g/mol) 208.345
MDL Number MFCD09750931
InChI Key ICCIOWLZPJIEBA-UHFFFAOYSA-N
PubChem CID 10987498
IUPAC Name 4-(4-ethylcyclohexyl)cyclohexan-1-one
SMILES CCC1CCC(CC1)C2CCC(=O)CC2

Specifications

Color Colorless
Quantity 5 g
Formula Weight 208.35
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-(trans-4-Ethylcyclohexyl)cyclohexanone
EINECSNumber (4)-1795
TSCA No