missing translation for 'onlineSavingsMsg'
Learn More

4-[trans-4-[(E)-1-Propenyl]cyclohexyl]benzonitrile 98.0+%, TCI America™

Catalog No. p23781g
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P23781G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P23781G Supplier TCI America Supplier No. P23781G

Chemical Identifiers

CAS 96184-40-6
Molecular Formula C16H19N
Molecular Weight (g/mol) 225.335
MDL Number MFCD24849961
InChI Key WFVBLRKVRNUULX-NSCUHMNNSA-N
Synonym 1-Cyano-4-[trans-4-[(E)-1-propenyl]cyclohexyl]benzene
PubChem CID 15606765
IUPAC Name 4-[4-[(E)-prop-1-enyl]cyclohexyl]benzonitrile
SMILES CC=CC1CCC(CC1)C2=CC=C(C=C2)C#N

Specifications

Melting Point 66°C
Color White
Quantity 1 g
UN Number 3439
Formula Weight 225.34
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-[trans-4-[(E)-1-Propenyl]cyclohexyl]benzonitrile