missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-(trans-4-Butylcyclohexyl)cyclohexanone 98.0+%, TCI America™
$46.20 - $169.41
Chemical Identifiers
| CAS | 92413-47-3 |
|---|---|
| Molecular Formula | C16H28O |
| Molecular Weight (g/mol) | 236.399 |
| MDL Number | MFCD06797514 |
| InChI Key | LDUYZTOYURUHIX-UHFFFAOYSA-N |
| PubChem CID | 23440378 |
| IUPAC Name | 4-(4-butylcyclohexyl)cyclohexan-1-one |
| SMILES | CCCCC1CCC(CC1)C2CCC(=O)CC2 |
Chemical Identifiers
| 92413-47-3 | |
| 236.399 | |
| LDUYZTOYURUHIX-UHFFFAOYSA-N | |
| 4-(4-butylcyclohexyl)cyclohexan-1-one |
| C16H28O | |
| MFCD06797514 | |
| 23440378 | |
| CCCCC1CCC(CC1)C2CCC(=O)CC2 |
Specifications
| 92413-47-3 | |
| White | |
| MFCD06797514 | |
| LDUYZTOYURUHIX-UHFFFAOYSA-N | |
| 4-(4-butylcyclohexyl)cyclohexan-1-one | |
| 23440378 | |
| ≥98.0% (GC) | |
| 4-(trans-4-Butylcyclohexyl)cyclohexanone |
| 44°C | |
| C16H28O | |
| 5 g | |
| CCCCC1CCC(CC1)C2CCC(=O)CC2 | |
| 236.399 | |
| 236.40 | |
| Crystalline Powder |
Safety and Handling
TSCA : No