missing translation for 'onlineSavingsMsg'
Learn More

4-(trans-4-Butylcyclohexyl)benzonitrile 98.0+%, TCI America™

Catalog No. B49245G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B49245G 5 g
B492425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B49245G Supplier TCI America Supplier No. B49245G

Chemical Identifiers

CAS 61204-00-0
Molecular Formula C17H23N
Molecular Weight (g/mol) 241.38
MDL Number MFCD00799425
InChI Key YYAVXASAKUOZJJ-UHFFFAOYSA-N
Synonym 4-trans-4-butylcyclohexyl benzonitrile, 4-4-butylcyclohexyl benzonitrile, p-4-butylcyclohexyl benzonitrile, benzonitrile, 4-4-butylcyclohexyl, benzonitrile, 4-trans-4-butylcyclohexyl, 4-trans-4-butylcyclohexyl-benzonitrile, trans-4-4'-n-butylcyclohexyl-benzonitrile, benzonitrile,4-trans-4-butylcyclohexyl, 4-4-butylcyclohexyl benzenecarbonitrile, 4-1s,4r-4-butylcyclohexyl benzonitrile
PubChem CID 109062
IUPAC Name 4-(4-butylcyclohexyl)benzonitrile
SMILES CCCCC1CCC(CC1)C1=CC=C(C=C1)C#N

Specifications

Melting Point 43°C
Color White
Quantity 5 g
UN Number 3439
Formula Weight 241.38
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-(trans-4-Butylcyclohexyl)benzonitrile