Learn More
4-Tolylboronic acid, 97%
CAS: 5720-05-8 | C7H9BO2 | 135.96 g/mol
$75.77 - $224.51
Chemical Identifiers
| CAS | 5720-05-8 |
|---|---|
| Molecular Formula | C7H9BO2 |
| Molecular Weight (g/mol) | 135.96 |
| MDL Number | MFCD00039138 |
| InChI Key | BIWQNIMLAISTBV-UHFFFAOYSA-N |
| Synonym | 4-tolylboronic acid, p-tolylboronic acid, 4-methylphenyl boronic acid, p-tolueneboronic acid, 4-methylbenzeneboronic acid, p-methylbenzeneboronic acid, boronic acid, 4-methylphenyl, boronic acid, p-tolyl, 4-methylphenylboric acid, tolyl boronic acid |
| PubChem CID | 79799 |
| IUPAC Name | (4-methylphenyl)boronic acid |
| SMILES | CC1=CC=C(C=C1)B(O)O |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC344430010
|
Thermo Scientific Chemicals
344430010 |
1 g | Glass bottle |
Each for $75.77
|
|
||||
|
AC344430050
|
Thermo Scientific Chemicals
344430050 |
5 g | Glass bottle |
Each for $224.51
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 5720-05-8 | |
| 135.96 | |
| BIWQNIMLAISTBV-UHFFFAOYSA-N | |
| 79799 | |
| CC1=CC=C(C=C1)B(O)O |
| C7H9BO2 | |
| MFCD00039138 | |
| 4-tolylboronic acid, p-tolylboronic acid, 4-methylphenyl boronic acid, p-tolueneboronic acid, 4-methylbenzeneboronic acid, p-methylbenzeneboronic acid, boronic acid, 4-methylphenyl, boronic acid, p-tolyl, 4-methylphenylboric acid, tolyl boronic acid | |
| (4-methylphenyl)boronic acid |
Specifications
| 5720-05-8 | |
| 100.0 | |
| White | |
| 97% | |
| C7H9BO2 | |
| MFCD00039138 | |
| 4-tolylboronic acid, p-tolylboronic acid, 4-methylphenyl boronic acid, p-tolueneboronic acid, 4-methylbenzeneboronic acid, p-methylbenzeneboronic acid, boronic acid, 4-methylphenyl, boronic acid, p-tolyl, 4-methylphenylboric acid, tolyl boronic acid | |
| CC1=CC=C(C=C1)B(O)O | |
| 135.96 | |
| 135.96 | |
| Crystalline Powder |
| 96.0 | |
| 256°C to 263°C | |
| Authentic | |
| Glass bottle | |
| CH3C6H4B(OH)2 | |
| 1 g | |
| BIWQNIMLAISTBV-UHFFFAOYSA-N | |
| (4-methylphenyl)boronic acid | |
| 79799 | |
| 97% | |
| 4-Tolylboronic acid |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Call a POISON CENTER or doctor/physicia
GHS Signal Word: Warning
RUO – Research Use Only