missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals 4-Thiouracil, 97%
$176.62 - $176.62
Chemical Identifiers
| CAS | 591-28-6 |
|---|---|
| Molecular Formula | C4H4N2OS |
| Molecular Weight (g/mol) | 128.15 |
| MDL Number | MFCD00090842 |
| InChI Key | OVONXEQGWXGFJD-UHFFFAOYSA-N |
| Synonym | 4-thiouracil, 2-hydroxy-4 1h-pyrimidinethione, 4-thioxo-3,4-dihydropyrimidin-2 1h-one, 2-hydroxy-4-mercaptopyrimidine, uracil, 4-thio, 4-mercaptopyrimidin-2-ol, 4-sulfanylpyrimidin-2-ol, 2-hydroxy-4-thiopyrimidine, 2 1h-pyrimidinone, 3,4-dihydro-4-thioxo, 2 1h-pyrimidinone, 4-mercapto-9ci |
| PubChem CID | 2734394 |
| IUPAC Name | 4-sulfanylidene-1H-pyrimidin-2-one |
| SMILES | O=C1NC=CC(=S)N1 |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC359930010
|
Thermo Scientific Chemicals
359930010 |
1 g | Glass bottle |
N/A
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 591-28-6 | |
| 128.15 | |
| OVONXEQGWXGFJD-UHFFFAOYSA-N | |
| 2734394 | |
| O=C1NC=CC(=S)N1 |
| C4H4N2OS | |
| MFCD00090842 | |
| 4-thiouracil, 2-hydroxy-4 1h-pyrimidinethione, 4-thioxo-3,4-dihydropyrimidin-2 1h-one, 2-hydroxy-4-mercaptopyrimidine, uracil, 4-thio, 4-mercaptopyrimidin-2-ol, 4-sulfanylpyrimidin-2-ol, 2-hydroxy-4-thiopyrimidine, 2 1h-pyrimidinone, 3,4-dihydro-4-thioxo, 2 1h-pyrimidinone, 4-mercapto-9ci | |
| 4-sulfanylidene-1H-pyrimidin-2-one |
Specifications
| 591-28-6 | |
| 100.0 | |
| Yellow | |
| 96% min. (HPLC) | |
| C4H4N2OS | |
| 1 g | |
| OVONXEQGWXGFJD-UHFFFAOYSA-N | |
| 4-sulfanylidene-1H-pyrimidin-2-one | |
| 2734394 | |
| 97% | |
| 4-Thiouracil, 97% |
| 96.0 | |
| 295°C | |
| Authentic | |
| Glass bottle | |
| MFCD00090842 | |
| 4-thiouracil, 2-hydroxy-4 1h-pyrimidinethione, 4-thioxo-3,4-dihydropyrimidin-2 1h-one, 2-hydroxy-4-mercaptopyrimidine, uracil, 4-thio, 4-mercaptopyrimidin-2-ol, 4-sulfanylpyrimidin-2-ol, 2-hydroxy-4-thiopyrimidine, 2 1h-pyrimidinone, 3,4-dihydro-4-thioxo, 2 1h-pyrimidinone, 4-mercapto-9ci | |
| O=C1NC=CC(=S)N1 | |
| 128.15 | |
| 128.15 | |
| Powder |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
GHS P Statement
Wash face,hands and any exposed skin thoroughly after handling.
Do not eat,drink or smoke when using this product.
GHS Signal Word: Warning
RUO – Research Use Only