missing translation for 'onlineSavingsMsg'
Learn More

4-(thien-2-ylmethyl)benzoyl chloride, Thermo Scientific™

Catalog No. p-4391480
Change view
Click to view available options
Quantity:
250mg
1g
5g
Packaging:
Amber Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC70602CB 250mg Amber Glass Bottle
CC70602DA 1g Amber Glass Bottle
CC70602DE 5g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. CC70602CB Supplier Thermo Scientific Chemicals Supplier No. CC70602CB

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1151512-19-4
Molecular Formula C12H9ClOS
Molecular Weight (g/mol) 236.71
InChI Key AOHWXUBTRWZCOZ-UHFFFAOYSA-N
Synonym 4-thien-2-ylmethyl benzoyl chloride, 4-2-thienylmethyl benzoyl chloride, 4-thiophen-2-ylmethyl benzoyl chloride, 4-thiophen-2-yl methyl benzoyl chloride
PubChem CID 45594296

Specifications

Packaging Amber Glass Bottle
Quantity 250mg
Formula Weight 236.72
Percent Purity 97%
Chemical Name or Material 4-(thien-2-ylmethyl)benzoyl chloride
Hazard Category GHS Signal Word: Danger