missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-(thien-2-ylmethyl)benzoyl chloride, Thermo Scientific™
Specifications
Specifications
| Packaging | Amber Glass Bottle |
| Quantity | 250mg |
| Formula Weight | 236.72 |
| Percent Purity | 97% |
| Chemical Name or Material | 4-(thien-2-ylmethyl)benzoyl chloride |
Safety and Handling
| Hazard Category | GHS Signal Word: Danger |