missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Thiazolecarboxylic acid, 97%, Thermo Scientific™
Chemical Identifiers
| CAS | 3973-08-8 |
|---|---|
| Molecular Formula | C4H3NO2S |
| Molecular Weight (g/mol) | 129.14 |
| InChI Key | QJVVNQLFPMBGLC-UHFFFAOYSA-N |
| Synonym | 1,1,4,4-tetraethoxy-2-butyne, 2-butyne,1,1,4,4-tetraethoxy |
| PubChem CID | 2724486 |
| IUPAC Name | 1,1,4,4-tetraethoxybut-2-yne |
| SMILES | CCOC(C#CC(OCC)OCC)OCC |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AC443970010
|
Thermo Scientific Chemicals
443970010 |
1g |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
AC443970050
|
Thermo Scientific Chemicals
443970050 |
5g |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
| 3973-08-8 | |
| 129.14 | |
| 1,1,4,4-tetraethoxy-2-butyne, 2-butyne,1,1,4,4-tetraethoxy | |
| 1,1,4,4-tetraethoxybut-2-yne |
| C4H3NO2S | |
| QJVVNQLFPMBGLC-UHFFFAOYSA-N | |
| 2724486 | |
| CCOC(C#CC(OCC)OCC)OCC |
Specifications
| 3973-08-8 | |
| Authentic | |
| Glass Bottle | |
| 1g | |
| HMVYYTRDXNKRBQ-UHFFFAOYSA-N | |
| 1,3-thiazole-4-carboxylic acid | |
| 304271 | |
| 129.14 | |
| 4-Thiazolecarboxylic acid |
| 3% max. | |
| 96% min. (HPLC) | |
| C4H3NO2S | |
| thiazole-4-carboxylic acid, 4-thiazolecarboxylic acid, 4-thiazolecarboxylicacid, 4-carboxythiazole, 4-carboxy-1,3-thiazole, thiazole 4-carboxylic acid, acmc-2097eb, thiazol-4-carboxylic, ksc225i1n | |
| C1=C(N=CS1)C(=O)O | |
| 129.14 | |
| CHEBI:46230 | |
| 97% |
Safety and Handling
GHS H Statement
May cause an allergic skin reaction.
Causes serious eye irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF ON SKIN:
GHS Signal Word: Warning
RUO â Research Use Only