missing translation for 'onlineSavingsMsg'
Learn More

4-Thiazolecarboxylic acid, 97%

Catalog No. AC443970010
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC443970010 1 g
AC443970050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC443970010 Supplier Thermo Scientific Chemicals Supplier No. 443970010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 3973-08-8 | C4H3NO2S | 129.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 3973-08-8
Molecular Formula C4H3NO2S
Molecular Weight (g/mol) 129.14
ChEBI CHEBI:46230

Specifications

Melting Point 195.0°C to 199.0°C
Loss on Drying 3% max.
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass Bottle
Quantity 1 g
Synonym thiazole-4-carboxylic acid, 4-thiazolecarboxylic acid, 4-thiazolecarboxylicacid, 4-carboxythiazole, 4-carboxy-1,3-thiazole, thiazole 4-carboxylic acid, acmc-2097eb, thiazol-4-carboxylic, ksc225i1n
InChI Key HMVYYTRDXNKRBQ-UHFFFAOYSA-N
SMILES C1=C(N=CS1)C(=O)O
IUPAC Name 1,3-thiazole-4-carboxylic acid
PubChem CID 304271
Formula Weight 129.14
Percent Purity 97%
Chemical Name or Material 4-Thiazolecarboxylic acid
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
May cause an allergic skin reaction.
Causes serious eye irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF ON SKIN:

RUO – Research Use Only