missing translation for 'onlineSavingsMsg'
Learn More

4-(Tetrahydropyran-4-yloxy)benzoic acid, 97%, Thermo Scientific™

$173.20 - $173.20

Chemical Identifiers

CAS 851048-51-6
Molecular Formula C12H14O4
Molecular Weight (g/mol) 222.24
MDL Number MFCD09025874
InChI Key MPNYLYRGNCZFFH-UHFFFAOYSA-N
PubChem CID 24229519
IUPAC Name 4-(oxan-4-yloxy)benzoic acid
SMILES C1COCCC1OC2=CC=C(C=C2)C(=O)O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
CC52401CB
SDS View Documents
Thermo Scientific Chemicals
CC52401CB
250 mg Amber Glass Bottle
Each for $173.20
Only null left
 
Description

Description

Specifications

Chemical Identifiers

851048-51-6
222.24
MPNYLYRGNCZFFH-UHFFFAOYSA-N
4-(oxan-4-yloxy)benzoic acid
C12H14O4
MFCD09025874
24229519
C1COCCC1OC2=CC=C(C=C2)C(=O)O

Specifications

851048-51-6
100.0
Amber Glass Bottle
MFCD09025874
4-tetrahydro-2h-pyran-4-yl oxy benzoic acid, 4-oxan-4-yloxy benzoic acid, 4-tetrahydropyran-4-yloxy benzoic acid, 4-oxan-4-yl oxy benzoic acid, 4-tetrahydro-2h-pyran-4-yloxy benzoic acid, 4-tetrahydro-2h-pyran-4-yl oxy benzoicacid, benzoic acid, 4-tetrahydro-2h-pyran-4-yl oxy, benzoic acid,4-tetrahydro-2h-pyran-4-yl oxy, 4-2h-3,4,5,6-tetrahydropyran-4-yloxy benzoic acid
C1COCCC1OC2=CC=C(C=C2)C(=O)O
222.24
222.24
4-(Tetrahydropyran-4-yloxy)benzoic acid
96.5
97%
C12H14O4
250 mg
MPNYLYRGNCZFFH-UHFFFAOYSA-N
4-(oxan-4-yloxy)benzoic acid
24229519
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Harmful if inhaled.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS
missing translation for 'documents'

missing translation for 'documents'