missing translation for 'onlineSavingsMsg'
Learn More

4-tert-Butylsulfonylcalix[4]arene 98.0+%, TCI America™

Catalog No. p-7119613
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B23121G 1 g
B23125G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B23121G Supplier TCI America Supplier No. B23121G
Only null left

Chemical Identifiers

CAS 204190-49-8
Molecular Formula C40H48O12S4
Molecular Weight (g/mol) 849.05
MDL Number MFCD06797060
InChI Key BOLKLFFOYGXSKT-UHFFFAOYSA-N
Synonym Tetra-tert-butyl(tetrahydroxy)tetrasulfonylcalix[4]arene
PubChem CID 11445832
IUPAC Name 5,11,17,23-tetra-tert-butyl-25,26,27,28-tetrahydroxy-2λ⁶,8λ⁶,14λ⁶,20λ⁶-tetrathiapentacyclo[19.3.1.1³,⁷.1⁹,¹³.1¹⁵,¹⁹]octacosa-1(25),3,5,7(28),9(27),10,12,15(26),16,18,21,23-dodecaen-2,2,8,8,14,14,20,20-octone
SMILES CC(C)(C)C1=CC2=C(O)C(=C1)S(=O)(=O)C1=CC(=CC(=C1O)S(=O)(=O)C1=CC(=CC(=C1O)S(=O)(=O)C1=C(O)C(=CC(=C1)C(C)(C)C)S2(=O)=O)C(C)(C)C)C(C)(C)C

Specifications

Quantity 1 g
Formula Weight 849.05
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 4-tert-Butylsulfonylcalix[4]arene