missing translation for 'onlineSavingsMsg'
Learn More

4'-[(tert-Butyldimethylsilyloxy)methyl]-2,2,2-trifluoroacetophenone 97.0+%, TCI America™

Catalog No. b51565g
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B51565G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B51565G Supplier TCI America Supplier No. B51565G
Only null left

Chemical Identifiers

CAS 87736-75-2
Molecular Formula C15H21F3O2Si
Molecular Weight (g/mol) 318.41
MDL Number MFCD29089349
InChI Key PNTKHPCHUVZRCB-UHFFFAOYSA-N
Synonym [(4-(Trifluoroacetyl)benzyl)oxy](tert-butyl)dimethylsilane
PubChem CID 13063196
IUPAC Name 1-(4-{[(tert-butyldimethylsilyl)oxy]methyl}phenyl)-2,2,2-trifluoroethan-1-one
SMILES CC(C)(C)[Si](C)(C)OCC1=CC=C(C=C1)C(=O)C(F)(F)F

Specifications

Color Yellow
Boiling Point 145°C
Quantity 5 g
Formula Weight 318.41
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 4′-[(tert-Butyldimethylsilyloxy)methyl]-2,2,2-trifluoroacetophenone