missing translation for 'onlineSavingsMsg'
Learn More

4-tert-Butylcyclohexylamine, cis + trans, 97%

Catalog No. aal0247214
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0247214 25 g
AAL0247206 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0247214 Supplier Thermo Scientific Chemicals Supplier No. L0247214
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 5400-88-4 | C10H21N | 155.29 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 5400-88-4
Molecular Formula C10H21N
Molecular Weight (g/mol) 155.29
MDL Number MFCD00013763,MFCD20690546
InChI Key BGNLXETYTAAURD-UHFFFAOYSA-N
Synonym 4-tert-butylcyclohexylamine, 4-tert-butyl cyclohexanamine, 4-t-butylcyclohexylamine, 4-tert-butylcyclohexanamine, 4-amino-1-tert-butylcyclohexane, 4-tert.-butylcyclohexylamine, 4-tert-butylcyclohexyl amine, cyclohexanamine, 4-1,1-dimethylethyl, cis-1-amino-4-tert-butylcyclohexane, 4-tert-butylcyclohexylamine,c&t
PubChem CID 79396
IUPAC Name 4-tert-butylcyclohexan-1-amine
SMILES CC(C)(C)C1CCC(N)CC1

Specifications

Density 0.87
Flash Point 79°C (174°F)
Refractive Index 1.4662
Quantity 25 g
UN Number UN2735
Beilstein 1523780
Sensitivity Air Sensitive
Formula Weight 155.29
Percent Purity 97%
Chemical Name or Material 4-tert-Butylcyclohexylamine, Cis + Trans
Hazard Category H227-H314-H335
Hazard Statement GHS H Statement
H314-H318-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.
Precautionary Statement P210-P235-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.
EINECSNumber 226-444-7
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only