missing translation for 'onlineSavingsMsg'
Learn More

4-tert-Butylcyclohexylamine, 97%, mixture of cis and trans, Thermo Scientific™

Catalog No. p-7077566
Change view
Click to view available options
Quantity:
5mL
25mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC455600050 5mL
AC455600250 25mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC455600050 Supplier Thermo Scientific Chemicals Supplier No. 455600050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Chemical Identifiers

CAS 5400-88-4
Molecular Formula C10H21N
Molecular Weight (g/mol) 155.29
MDL Number MFCD00013763,MFCD20690546
InChI Key BGNLXETYTAAURD-UHFFFAOYSA-N
Synonym 4-tert-butylcyclohexylamine, 4-tert-butyl cyclohexanamine, 4-t-butylcyclohexylamine, 4-tert-butylcyclohexanamine, 4-amino-1-tert-butylcyclohexane, 4-tert.-butylcyclohexylamine, 4-tert-butylcyclohexyl amine, cyclohexanamine, 4-1,1-dimethylethyl, cis-1-amino-4-tert-butylcyclohexane, 4-tert-butylcyclohexylamine,c&t
PubChem CID 79396
IUPAC Name 4-tert-butylcyclohexan-1-amine
SMILES CC(C)(C)C1CCC(N)CC1

Specifications

Density 0.8700g/mL
Flash Point 79°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (total of isomers) (GC)
Quantity 5mL
Specific Gravity 0.87
Solubility Information Solubility in water: immiscible.
Formula Weight 155.29
Percent Purity 97%
Chemical Name or Material 4-tert-Butylcyclohexylamine
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately
EINECSNumber 226-444-7

RUO – Research Use Only