missing translation for 'onlineSavingsMsg'
Learn More

4-tert-Butylcyclohexylamine, 97%, mixture of cis and trans, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 455600250

Quantity 25mL

 View more versions of this product

Catalog No. AC455600250

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

5400-88-4
155.29
BGNLXETYTAAURD-UHFFFAOYSA-N
79396
CC(C)(C)C1CCC(N)CC1
C10H21N
MFCD00013763,MFCD20690546
4-tert-butylcyclohexylamine, 4-tert-butyl cyclohexanamine, 4-t-butylcyclohexylamine, 4-tert-butylcyclohexanamine, 4-amino-1-tert-butylcyclohexane, 4-tert.-butylcyclohexylamine, 4-tert-butylcyclohexyl amine, cyclohexanamine, 4-1,1-dimethylethyl, cis-1-amino-4-tert-butylcyclohexane, 4-tert-butylcyclohexylamine,c&t
4-tert-butylcyclohexan-1-amine

Specifications

4-tert-Butylcyclohexylamine
0.8700g/mL
Authentic
C10H21N
25mL
4-tert-butylcyclohexylamine, 4-tert-butyl cyclohexanamine, 4-t-butylcyclohexylamine, 4-tert-butylcyclohexanamine, 4-amino-1-tert-butylcyclohexane, 4-tert.-butylcyclohexylamine, 4-tert-butylcyclohexyl amine, cyclohexanamine, 4-1,1-dimethylethyl, cis-1-amino-4-tert-butylcyclohexane, 4-tert-butylcyclohexylamine,c&t
BGNLXETYTAAURD-UHFFFAOYSA-N
4-tert-butylcyclohexan-1-amine
79396
97%
5400-88-4
79°C
96% min. (total of isomers) (GC)
MFCD00013763,MFCD20690546
0.87
Solubility in water: immiscible.
CC(C)(C)C1CCC(N)CC1
155.29
155.29
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately

GHS Signal Word: Danger

EINECSNumber : 226-444-7

SDS
Documents

Documents

Promotions

Promotions

Promo
Expires: 06-30-2027
Get Up to $1,800 in Chemicals with Purchase
Spend $250 or more on Thermo Fisher Scientific chemicals and get up to $1,800 to spend on additional chemicals.

Adding...