missing translation for 'onlineSavingsMsg'
Learn More

4-tert-Butylcyclohexanol, 99%, mixture of isomers

Catalog No. p-79905
Change view
Click to view available options
Quantity:
5 g
250 g
Packaging:
Glass Bottle
Plastic bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC107900050 5 g Glass Bottle
AC107902500 250 g Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC107900050 Supplier Thermo Scientific Chemicals Supplier No. 107900050
Only null left

CAS: 98-52-2 | C10H20O | 156.27 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 98-52-2
Molecular Formula C10H20O
Molecular Weight (g/mol) 156.27
MDL Number MFCD00001473,MFCD00064952,MFCD00070476
InChI Key CCOQPGVQAWPUPE-UHFFFAOYSA-N
Synonym 4-tert-butylcyclohexanol, cis-4-tert-butylcyclohexanol, trans-4-tert-butylcyclohexanol, 4-tert-butyl cyclohexanol, cis-4-tert-butyl cyclohexanol, padaryl, p-tert-butylcyclohexanol, cyclohexanol, 4-1,1-dimethylethyl, cyclohexanol, 4-tert-butyl, 4-t-butylcyclohexanol
PubChem CID 7391
IUPAC Name 4-tert-butylcyclohexan-1-ol
SMILES CC(C)(C)C1CCC(O)CC1

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 60.0°C to 69.0°C
Color White
Boiling Point 224.0°C
Flash Point 200°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass Bottle
Refractive Index 1.447
Linear Formula (CH3)3CC6H10OH
Quantity 5 g
Beilstein 06, I, 18
Solubility Information Solubility in water: <1g/L water (20 c)
Viscosity 8.68 mPa.s (80°C)
Formula Weight 156.27
Percent Purity 99%
Physical Form Powder or Granules
Chemical Name or Material 4-tert-Butylcyclohexanol, 99%
Show More Show Less
Hazard Category GHS Signal Word: Warning
EINECSNumber 202-676-4
RTECSNumber GV8750000
TSCA TSCA

RUO – Research Use Only