missing translation for 'onlineSavingsMsg'
Learn More

4-tert-Butylcalix[8]arene 98.0+%, TCI America™

Catalog No. B17505G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B17505G 5 g
B175025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B17505G Supplier TCI America Supplier No. B17505G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 68971-82-4
Molecular Formula C88H112O8
Molecular Weight (g/mol) 1297.86
MDL Number MFCD00075392
InChI Key OLZFZIXORGGLLS-UHFFFAOYSA-N
Synonym 4-tert-butylcalix 8 arene, p-t-butylcalix 8 arene, p-tert-butylcalix 8 arene, ncistruc1_000782, ncistruc2_001502, 3,3-dimethyl-2-epoxybutane, 4-tert-butyl calix 8 arene, octa-tert-butyl octahydroxy calix 8 arene
PubChem CID 434172
IUPAC Name 5,11,17,23,29,35,41,47-octa-tert-butylnonacyclo[43.3.1.1³,⁷.1⁹,¹³.1¹⁵,¹⁹.1²¹,²⁵.1²⁷,³¹.1³³,³⁷.1³⁹,⁴³]hexapentaconta-1(49),3,5,7(56),9,11,13(55),15,17,19(54),21,23,25(53),27,29,31(52),33,35,37(51),39,41,43(50),45,47-tetracosaen-49,50,51,52,53,54,55,56-octol
SMILES CC(C)(C)C1=CC2=C(O)C(CC3=C(O)C(CC4=C(O)C(CC5=C(O)C(CC6=C(O)C(CC7=C(O)C(CC8=C(O)C(CC9=C(O)C(C2)=CC(=C9)C(C)(C)C)=CC(=C8)C(C)(C)C)=CC(=C7)C(C)(C)C)=CC(=C6)C(C)(C)C)=CC(=C5)C(C)(C)C)=CC(=C4)C(C)(C)C)=CC(=C3)C(C)(C)C)=C1

Specifications

Color White
Quantity 5 g
Formula Weight 1297.86
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 4-tert-Butylcalix[8]arene