missing translation for 'onlineSavingsMsg'
Learn More

4-tert-Butylcalix[8]arene, 96%

CAS: 68971-82-4 | C88H112O8 | 1297.86 g/mol

$76.26 - $879.12

Chemical Identifiers

CAS 68971-82-4
Molecular Formula C88H112O8
Molecular Weight (g/mol) 1297.86
MDL Number MFCD00075392
InChI Key OLZFZIXORGGLLS-UHFFFAOYSA-N
Synonym 4-tert-butylcalix 8 arene, p-t-butylcalix 8 arene, p-tert-butylcalix 8 arene, ncistruc1_000782, ncistruc2_001502, 3,3-dimethyl-2-epoxybutane, 4-tert-butyl calix 8 arene, octa-tert-butyl octahydroxy calix 8 arene
PubChem CID 434172
IUPAC Name 5,11,17,23,29,35,41,47-octa-tert-butylnonacyclo[43.3.1.1³,⁷.1⁹,¹³.1¹⁵,¹⁹.1²¹,²⁵.1²⁷,³¹.1³³,³⁷.1³⁹,⁴³]hexapentaconta-1(49),3,5,7(56),9,11,13(55),15,17,19(54),21,23,25(53),27,29,31(52),33,35,37(51),39,41,43(50),45,47-tetracosaen-49,50,51,52,53,54,55,56-octol
SMILES CC(C)(C)C1=CC2=C(O)C(CC3=C(O)C(CC4=C(O)C(CC5=C(O)C(CC6=C(O)C(CC7=C(O)C(CC8=C(O)C(CC9=C(O)C(C2)=CC(=C9)C(C)(C)C)=CC(=C8)C(C)(C)C)=CC(=C7)C(C)(C)C)=CC(=C6)C(C)(C)C)=CC(=C5)C(C)(C)C)=CC(=C4)C(C)(C)C)=CC(=C3)C(C)(C)C)=C1
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAB2144303
View Documents
Thermo Scientific Chemicals
B2144303
1 g
Each for $76.26
Only null left
 
AAB2144306
View Documents
Thermo Scientific Chemicals
B2144306
5 g
Each for $257.74
Only null left
 
AAB2144314
View Documents
Thermo Scientific Chemicals
B2144314
25 g
Each for $879.12
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

68971-82-4
1297.86
OLZFZIXORGGLLS-UHFFFAOYSA-N
434172
CC(C)(C)C1=CC2=C(O)C(CC3=C(O)C(CC4=C(O)C(CC5=C(O)C(CC6=C(O)C(CC7=C(O)C(CC8=C(O)C(CC9=C(O)C(C2)=CC(=C9)C(C)(C)C)=CC(=C8)C(C)(C)C)=CC(=C7)C(C)(C)C)=CC(=C6)C(C)(C)C)=CC(=C5)C(C)(C)C)=CC(=C4)C(C)(C)C)=CC(=C3)C(C)(C)C)=C1
C88H112O8
MFCD00075392
4-tert-butylcalix 8 arene, p-t-butylcalix 8 arene, p-tert-butylcalix 8 arene, ncistruc1_000782, ncistruc2_001502, 3,3-dimethyl-2-epoxybutane, 4-tert-butyl calix 8 arene, octa-tert-butyl octahydroxy calix 8 arene
5,11,17,23,29,35,41,47-octa-tert-butylnonacyclo[43.3.1.1³,⁷.1⁹,¹³.1¹⁵,¹⁹.1²¹,²⁵.1²⁷,³¹.1³³,³⁷.1³⁹,⁴³]hexapentaconta-1(49),3,5,7(56),9,11,13(55),15,17,19(54),21,23,25(53),27,29,31(52),33,35,37(51),39,41,43(50),45,47-tetracosaen-49,50,51,52,53,54,55,56-octol

Specifications

68971-82-4
C88H112O8
1 g
4-tert-butylcalix 8 arene, p-t-butylcalix 8 arene, p-tert-butylcalix 8 arene, ncistruc1_000782, ncistruc2_001502, 3,3-dimethyl-2-epoxybutane, 4-tert-butyl calix 8 arene, octa-tert-butyl octahydroxy calix 8 arene
CC(C)(C)C1=CC2=C(O)C(CC3=C(O)C(CC4=C(O)C(CC5=C(O)C(CC6=C(O)C(CC7=C(O)C(CC8=C(O)C(CC9=C(O)C(C2)=CC(=C9)C(C)(C)C)=CC(=C8)C(C)(C)C)=CC(=C7)C(C)(C)C)=CC(=C6)C(C)(C)C)=CC(=C5)C(C)(C)C)=CC(=C4)C(C)(C)C)=CC(=C3)C(C)(C)C)=C1
1297.86
1297.87
4-tert-Butylcalix[8]arene
411°C to 412°C
MFCD00075392
2612052
OLZFZIXORGGLLS-UHFFFAOYSA-N
5,11,17,23,29,35,41,47-octa-tert-butylnonacyclo[43.3.1.1³,⁷.1⁹,¹³.1¹⁵,¹⁹.1²¹,²⁵.1²⁷,³¹.1³³,³⁷.1³⁹,⁴³]hexapentaconta-1(49),3,5,7(56),9,11,13(55),15,17,19(54),21,23,25(53),27,29,31(52),33,35,37(51),39,41,43(50),45,47-tetracosaen-49,50,51,52,53,54,55,56-octol
434172
96%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only