missing translation for 'onlineSavingsMsg'
Learn More

4-tert-Butylcalix[8]arene, 96%

Catalog No. AAB2144314
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2144314 25 g
AAB2144303 1 g
AAB2144306 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2144314 Supplier Thermo Scientific Chemicals Supplier No. B2144314
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 68971-82-4 | C88H112O8 | 1297.86 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 68971-82-4
Molecular Formula C88H112O8
Molecular Weight (g/mol) 1297.86
MDL Number MFCD00075392
InChI Key OLZFZIXORGGLLS-UHFFFAOYSA-N
Synonym 4-tert-butylcalix 8 arene, p-t-butylcalix 8 arene, p-tert-butylcalix 8 arene, ncistruc1_000782, ncistruc2_001502, 3,3-dimethyl-2-epoxybutane, 4-tert-butyl calix 8 arene, octa-tert-butyl octahydroxy calix 8 arene
PubChem CID 434172
IUPAC Name 5,11,17,23,29,35,41,47-octa-tert-butylnonacyclo[43.3.1.1³,⁷.1⁹,¹³.1¹⁵,¹⁹.1²¹,²⁵.1²⁷,³¹.1³³,³⁷.1³⁹,⁴³]hexapentaconta-1(49),3,5,7(56),9,11,13(55),15,17,19(54),21,23,25(53),27,29,31(52),33,35,37(51),39,41,43(50),45,47-tetracosaen-49,50,51,52,53,54,55,56-octol
SMILES CC(C)(C)C1=CC2=C(O)C(CC3=C(O)C(CC4=C(O)C(CC5=C(O)C(CC6=C(O)C(CC7=C(O)C(CC8=C(O)C(CC9=C(O)C(C2)=CC(=C9)C(C)(C)C)=CC(=C8)C(C)(C)C)=CC(=C7)C(C)(C)C)=CC(=C6)C(C)(C)C)=CC(=C5)C(C)(C)C)=CC(=C4)C(C)(C)C)=CC(=C3)C(C)(C)C)=C1

Specifications

Melting Point 411°C to 412°C
Quantity 25 g
Beilstein 2612052
Formula Weight 1297.87
Percent Purity 96%
Chemical Name or Material 4-tert-Butylcalix[8]arene
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only