missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-tert-Butylcalix[4]arene 98.0+%, TCI America™
$111.84 - $954.40
Chemical Identifiers
| CAS | 60705-62-6 |
|---|---|
| Molecular Formula | C44H56O4 |
| Molecular Weight (g/mol) | 648.93 |
| MDL Number | MFCD00066280 |
| InChI Key | NVKLTRSBZLYZHK-UHFFFAOYSA-N |
| Synonym | 4-tert-butylcalix 4 arene, tetra-tert-butyl tetrahydroxy calix 4 arene, 5,11,17,23-tetra-t-butyl-25,26,27,28-tetrahydroxycalix-4-arene, 4-t-butylcalix 4 arene, tetratert-butyl ? tetrol, p-tert-butylcalix 4 arene, 4-tert-butylcalix 4 arene 5g, ethyl 2-5,11,17,23-tetra tert-butyl-26,27,28-tris 2-ethoxy-2-oxyethoxy pentacyclo 19.3.1.1∼3,7∼.1∼9,13∼.1∼15,19∼ octacosa-1 25 ,3 28 ,4,6,9 27 ,10,12,15,17, |
| PubChem CID | 335377 |
| IUPAC Name | 5,11,17,23-tetra-tert-butylpentacyclo[19.3.1.1³,â·.1â¹,¹³.1¹âµ,¹â¹]octacosa-1(25),3,5,7(28),9,11,13(27),15,17,19(26),21,23-dodecaene-25,26,27,28-tetrol |
| SMILES | CC(C)(C)C1=CC2=C(O)C(CC3=C(O)C(CC4=C(O)C(CC5=C(O)C(C2)=CC(=C5)C(C)(C)C)=CC(=C4)C(C)(C)C)=CC(=C3)C(C)(C)C)=C1 |
Chemical Identifiers
| 60705-62-6 | |
| 648.93 | |
| NVKLTRSBZLYZHK-UHFFFAOYSA-N | |
| 335377 | |
| CC(C)(C)C1=CC2=C(O)C(CC3=C(O)C(CC4=C(O)C(CC5=C(O)C(C2)=CC(=C5)C(C)(C)C)=CC(=C4)C(C)(C)C)=CC(=C3)C(C)(C)C)=C1 |
| C44H56O4 | |
| MFCD00066280 | |
| 4-tert-butylcalix 4 arene, tetra-tert-butyl tetrahydroxy calix 4 arene, 5,11,17,23-tetra-t-butyl-25,26,27,28-tetrahydroxycalix-4-arene, 4-t-butylcalix 4 arene, tetratert-butyl ? tetrol, p-tert-butylcalix 4 arene, 4-tert-butylcalix 4 arene 5g, ethyl 2-5,11,17,23-tetra tert-butyl-26,27,28-tris 2-ethoxy-2-oxyethoxy pentacyclo 19.3.1.1∼3,7∼.1∼9,13∼.1∼15,19∼ octacosa-1 25 ,3 28 ,4,6,9 27 ,10,12,15,17, | |
| 5,11,17,23-tetra-tert-butylpentacyclo[19.3.1.1³,â·.1â¹,¹³.1¹âµ,¹â¹]octacosa-1(25),3,5,7(28),9,11,13(27),15,17,19(26),21,23-dodecaene-25,26,27,28-tetrol |
Specifications
| 60705-62-6 | |
| C44H56O4 | |
| 1 g | |
| NVKLTRSBZLYZHK-UHFFFAOYSA-N | |
| 5,11,17,23-tetra-tert-butylpentacyclo[19.3.1.1³,â·.1â¹,¹³.1¹âµ,¹â¹]octacosa-1(25),3,5,7(28),9,11,13(27),15,17,19(26),21,23-dodecaene-25,26,27,28-tetrol | |
| 335377 | |
| ≥98.0% (HPLC) | |
| 4-tert-Butylcalix[4]arene |
| White-Yellow | |
| MFCD00066280 | |
| 4-tert-butylcalix 4 arene, tetra-tert-butyl tetrahydroxy calix 4 arene, 5,11,17,23-tetra-t-butyl-25,26,27,28-tetrahydroxycalix-4-arene, 4-t-butylcalix 4 arene, tetratert-butyl ? tetrol, p-tert-butylcalix 4 arene, 4-tert-butylcalix 4 arene 5g, ethyl 2-5,11,17,23-tetra tert-butyl-26,27,28-tris 2-ethoxy-2-oxyethoxy pentacyclo 19.3.1.1∼3,7∼.1∼9,13∼.1∼15,19∼ octacosa-1 25 ,3 28 ,4,6,9 27 ,10,12,15,17, | |
| CC(C)(C)C1=CC2=C(O)C(CC3=C(O)C(CC4=C(O)C(CC5=C(O)C(C2)=CC(=C5)C(C)(C)C)=CC(=C4)C(C)(C)C)=CC(=C3)C(C)(C)C)=C1 | |
| 648.93 | |
| 648.93 | |
| Crystalline Powder |
Safety and Handling
TSCA : No