missing translation for 'onlineSavingsMsg'
Learn More

4-tert-Butylcalix[4]arene, 99%

CAS: 60705-62-6 | C44H56O4 | 648.93 g/mol

$206.53 - $743.19

Chemical Identifiers

CAS 60705-62-6
Molecular Formula C44H56O4
Molecular Weight (g/mol) 648.93
MDL Number MFCD00066280
InChI Key NVKLTRSBZLYZHK-UHFFFAOYSA-N
Synonym 4-tert-butylcalix 4 arene, tetra-tert-butyl tetrahydroxy calix 4 arene, 5,11,17,23-tetra-t-butyl-25,26,27,28-tetrahydroxycalix-4-arene, 4-t-butylcalix 4 arene, tetratert-butyl ? tetrol, p-tert-butylcalix 4 arene, 4-tert-butylcalix 4 arene 5g, ethyl 2-5,11,17,23-tetra tert-butyl-26,27,28-tris 2-ethoxy-2-oxyethoxy pentacyclo 19.3.1.1∼3,7∼.1∼9,13∼.1∼15,19∼ octacosa-1 25 ,3 28 ,4,6,9 27 ,10,12,15,17,
PubChem CID 335377
IUPAC Name 5,11,17,23-tetra-tert-butylpentacyclo[19.3.1.1³,⁷.1⁹,¹³.1¹⁵,¹⁹]octacosa-1(25),3,5,7(28),9,11,13(27),15,17,19(26),21,23-dodecaene-25,26,27,28-tetrol
SMILES CC(C)(C)C1=CC2=C(O)C(CC3=C(O)C(CC4=C(O)C(CC5=C(O)C(C2)=CC(=C5)C(C)(C)C)=CC(=C4)C(C)(C)C)=CC(=C3)C(C)(C)C)=C1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAB2151506
View Documents
Thermo Scientific Chemicals
B2151506
5 g
Each for $206.53
Only null left
 
AAB2151514
View Documents
Thermo Scientific Chemicals
B2151514
25 g
Each for $743.19
Only null left
 
Description

Description

It is used as a starting material for further derivatization, including halogenation, acylation, and diazo coupling.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as a starting material for further derivatization, including halogenation, acylation, and diazo coupling.

Solubility
Soluble in chloroform, benzene, toluene. Insoluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Specifications

Chemical Identifiers

60705-62-6
648.93
NVKLTRSBZLYZHK-UHFFFAOYSA-N
335377
CC(C)(C)C1=CC2=C(O)C(CC3=C(O)C(CC4=C(O)C(CC5=C(O)C(C2)=CC(=C5)C(C)(C)C)=CC(=C4)C(C)(C)C)=CC(=C3)C(C)(C)C)=C1
C44H56O4
MFCD00066280
4-tert-butylcalix 4 arene, tetra-tert-butyl tetrahydroxy calix 4 arene, 5,11,17,23-tetra-t-butyl-25,26,27,28-tetrahydroxycalix-4-arene, 4-t-butylcalix 4 arene, tetratert-butyl ? tetrol, p-tert-butylcalix 4 arene, 4-tert-butylcalix 4 arene 5g, ethyl 2-5,11,17,23-tetra tert-butyl-26,27,28-tris 2-ethoxy-2-oxyethoxy pentacyclo 19.3.1.1∼3,7∼.1∼9,13∼.1∼15,19∼ octacosa-1 25 ,3 28 ,4,6,9 27 ,10,12,15,17,
5,11,17,23-tetra-tert-butylpentacyclo[19.3.1.1³,⁷.1⁹,¹³.1¹⁵,¹⁹]octacosa-1(25),3,5,7(28),9,11,13(27),15,17,19(26),21,23-dodecaene-25,26,27,28-tetrol

Specifications

60705-62-6
C44H56O4
5 g
4-tert-butylcalix 4 arene, tetra-tert-butyl tetrahydroxy calix 4 arene, 5,11,17,23-tetra-t-butyl-25,26,27,28-tetrahydroxycalix-4-arene, 4-t-butylcalix 4 arene, tetratert-butyl ? tetrol, p-tert-butylcalix 4 arene, 4-tert-butylcalix 4 arene 5g, ethyl 2-5,11,17,23-tetra tert-butyl-26,27,28-tris 2-ethoxy-2-oxyethoxy pentacyclo 19.3.1.1∼3,7∼.1∼9,13∼.1∼15,19∼ octacosa-1 25 ,3 28 ,4,6,9 27 ,10,12,15,17,
NVKLTRSBZLYZHK-UHFFFAOYSA-N
5,11,17,23-tetra-tert-butylpentacyclo[19.3.1.1³,⁷.1⁹,¹³.1¹⁵,¹⁹]octacosa-1(25),3,5,7(28),9,11,13(27),15,17,19(26),21,23-dodecaene-25,26,27,28-tetrol
335377
99%
339°C to 344°C
MFCD00066280
2033965
Soluble in chloroform,benzene,toluene. Insoluble in water.
CC(C)(C)C1=CC2=C(O)C(CC3=C(O)C(CC4=C(O)C(CC5=C(O)C(C2)=CC(=C5)C(C)(C)C)=CC(=C4)C(C)(C)C)=CC(=C3)C(C)(C)C)=C1
648.93
648.93
4-tert-Butylcalix[4]arene
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 000-000-0

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only