missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-tert-Butylcalix[4]arene, 99%
CAS: 60705-62-6 | C44H56O4 | 648.93 g/mol
$794.84 - $794.84
Chemical Identifiers
| CAS | 60705-62-6 |
|---|---|
| Molecular Formula | C44H56O4 |
| Molecular Weight (g/mol) | 648.93 |
| MDL Number | MFCD00066280 |
| InChI Key | NVKLTRSBZLYZHK-UHFFFAOYSA-N |
| Synonym | 4-tert-butylcalix 4 arene, tetra-tert-butyl tetrahydroxy calix 4 arene, 5,11,17,23-tetra-t-butyl-25,26,27,28-tetrahydroxycalix-4-arene, 4-t-butylcalix 4 arene, tetratert-butyl ? tetrol, p-tert-butylcalix 4 arene, 4-tert-butylcalix 4 arene 5g, ethyl 2-5,11,17,23-tetra tert-butyl-26,27,28-tris 2-ethoxy-2-oxyethoxy pentacyclo 19.3.1.1∼3,7∼.1∼9,13∼.1∼15,19∼ octacosa-1 25 ,3 28 ,4,6,9 27 ,10,12,15,17, |
| PubChem CID | 335377 |
| IUPAC Name | 5,11,17,23-tetra-tert-butylpentacyclo[19.3.1.1³,â·.1â¹,¹³.1¹âµ,¹â¹]octacosa-1(25),3,5,7(28),9,11,13(27),15,17,19(26),21,23-dodecaene-25,26,27,28-tetrol |
| SMILES | CC(C)(C)C1=CC2=C(O)C(CC3=C(O)C(CC4=C(O)C(CC5=C(O)C(C2)=CC(=C5)C(C)(C)C)=CC(=C4)C(C)(C)C)=CC(=C3)C(C)(C)C)=C1 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AC290220250
|
Thermo Scientific Chemicals
290220250 |
25 g |
N/A
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chelation/Complexation reagentChemical Identifiers
| 60705-62-6 | |
| 648.93 | |
| NVKLTRSBZLYZHK-UHFFFAOYSA-N | |
| 335377 | |
| CC(C)(C)C1=CC2=C(O)C(CC3=C(O)C(CC4=C(O)C(CC5=C(O)C(C2)=CC(=C5)C(C)(C)C)=CC(=C4)C(C)(C)C)=CC(=C3)C(C)(C)C)=C1 |
| C44H56O4 | |
| MFCD00066280 | |
| 4-tert-butylcalix 4 arene, tetra-tert-butyl tetrahydroxy calix 4 arene, 5,11,17,23-tetra-t-butyl-25,26,27,28-tetrahydroxycalix-4-arene, 4-t-butylcalix 4 arene, tetratert-butyl ? tetrol, p-tert-butylcalix 4 arene, 4-tert-butylcalix 4 arene 5g, ethyl 2-5,11,17,23-tetra tert-butyl-26,27,28-tris 2-ethoxy-2-oxyethoxy pentacyclo 19.3.1.1∼3,7∼.1∼9,13∼.1∼15,19∼ octacosa-1 25 ,3 28 ,4,6,9 27 ,10,12,15,17, | |
| 5,11,17,23-tetra-tert-butylpentacyclo[19.3.1.1³,â·.1â¹,¹³.1¹âµ,¹â¹]octacosa-1(25),3,5,7(28),9,11,13(27),15,17,19(26),21,23-dodecaene-25,26,27,28-tetrol |
Specifications
| 60705-62-6 | |
| 100.0 | |
| White to Beige | |
| 98.5% min. (HPLC) | |
| C44H56O4 | |
| 25 g | |
| 4-tert-butylcalix 4 arene, tetra-tert-butyl tetrahydroxy calix 4 arene, 5,11,17,23-tetra-t-butyl-25,26,27,28-tetrahydroxycalix-4-arene, 4-t-butylcalix 4 arene, tetratert-butyl ? tetrol, p-tert-butylcalix 4 arene, 4-tert-butylcalix 4 arene 5g, ethyl 2-5,11,17,23-tetra tert-butyl-26,27,28-tris 2-ethoxy-2-oxyethoxy pentacyclo 19.3.1.1∼3,7∼.1∼9,13∼.1∼15,19∼ octacosa-1 25 ,3 28 ,4,6,9 27 ,10,12,15,17, | |
| NVKLTRSBZLYZHK-UHFFFAOYSA-N | |
| 5,11,17,23-tetra-tert-butylpentacyclo[19.3.1.1³,â·.1â¹,¹³.1¹âµ,¹â¹]octacosa-1(25),3,5,7(28),9,11,13(27),15,17,19(26),21,23-dodecaene-25,26,27,28-tetrol | |
| 335377 | |
| 99% | |
| 4-tert-Butylcalix[4]arene |
| 98.5 | |
| >300.0°C | |
| Authentic | |
| Glass bottle | |
| MFCD00066280 | |
| 06, IV, 7858 | |
| Solubility in water: insoluble. Other solubilities: soluble in chloroform,benzene,toluene | |
| CC(C)(C)C1=CC2=C(O)C(CC3=C(O)C(CC4=C(O)C(CC5=C(O)C(C2)=CC(=C5)C(C)(C)C)=CC(=C4)C(C)(C)C)=CC(=C3)C(C)(C)C)=C1 | |
| 648.93 | |
| 648.92 | |
| Crystalline Powder or Flakes |
RUO – Research Use Only