missing translation for 'onlineSavingsMsg'
Learn More

4-tert-Butylcalix[4]arene, 99%

Catalog No. AC290220250
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC290220250 25 g
AC290220050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC290220250 Supplier Thermo Scientific Chemicals Supplier No. 290220250
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 60705-62-6 | C44H56O4 | 648.93 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chelation/Complexation reagent

Chemical Identifiers

CAS 60705-62-6
Molecular Formula C44H56O4
Molecular Weight (g/mol) 648.93
MDL Number MFCD00066280
InChI Key NVKLTRSBZLYZHK-UHFFFAOYSA-N
Synonym 4-tert-butylcalix 4 arene, tetra-tert-butyl tetrahydroxy calix 4 arene, 5,11,17,23-tetra-t-butyl-25,26,27,28-tetrahydroxycalix-4-arene, 4-t-butylcalix 4 arene, tetratert-butyl ? tetrol, p-tert-butylcalix 4 arene, 4-tert-butylcalix 4 arene 5g, ethyl 2-5,11,17,23-tetra tert-butyl-26,27,28-tris 2-ethoxy-2-oxyethoxy pentacyclo 19.3.1.1∼3,7∼.1∼9,13∼.1∼15,19∼ octacosa-1 25 ,3 28 ,4,6,9 27 ,10,12,15,17,
PubChem CID 335377
IUPAC Name 5,11,17,23-tetra-tert-butylpentacyclo[19.3.1.1³,⁷.1⁹,¹³.1¹⁵,¹⁹]octacosa-1(25),3,5,7(28),9,11,13(27),15,17,19(26),21,23-dodecaene-25,26,27,28-tetrol
SMILES CC(C)(C)C1=CC2=C(O)C(CC3=C(O)C(CC4=C(O)C(CC5=C(O)C(C2)=CC(=C5)C(C)(C)C)=CC(=C4)C(C)(C)C)=CC(=C3)C(C)(C)C)=C1

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point >300.0°C
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (HPLC)
Packaging Glass bottle
Quantity 25 g
Beilstein 06, IV, 7858
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in chloroform,benzene,toluene
Formula Weight 648.92
Percent Purity 99%
Physical Form Crystalline Powder or Flakes
Chemical Name or Material 4-tert-Butylcalix[4]arene
Show More Show Less

RUO – Research Use Only