missing translation for 'onlineSavingsMsg'
Learn More

4-tert-Butylbiphenyl 98.0+%, TCI America™

Catalog No. B18115G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B18115G 5 g
B181125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B18115G Supplier TCI America Supplier No. B18115G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 1625-92-9
Molecular Formula C16H18
Molecular Weight (g/mol) 210.32
MDL Number MFCD00222366
InChI Key CDOYZTOFTGTGBC-UHFFFAOYSA-N
PubChem CID 137120
IUPAC Name 1-tert-butyl-4-phenylbenzene
SMILES CC(C)(C)C1=CC=C(C=C1)C2=CC=CC=C2

Specifications

Melting Point 52°C
Color White
Boiling Point 310°C
Quantity 5 g
Formula Weight 210.32
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-tert-Butylbiphenyl