missing translation for 'onlineSavingsMsg'
Learn More

4-tert-Butylbenzoyl chloride, 98%

Catalog No. p-78924
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC169100050 5 g Glass bottle
AC169100250 25 g Glass bottle
AC169101000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC169100050 Supplier Thermo Scientific Chemicals Supplier No. 169100050
Only null left

CAS: 1710-98-1 | C11H13ClO | 196.67 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1710-98-1
Molecular Formula C11H13ClO
Molecular Weight (g/mol) 196.67
MDL Number MFCD00000695
InChI Key WNLMYNASWOULQY-UHFFFAOYSA-N
PubChem CID 74372
IUPAC Name 4-tert-butylbenzoyl chloride
SMILES CC(C)(C)C1=CC=C(C=C1)C(Cl)=O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Colorless to Yellow
Density 1.0000g/mL
Boiling Point 135°C (20.0 mmHg)
Flash Point 87°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Refractive Index 1.5354 to 1.5374
Linear Formula (CH3)3CC6H4COCl
Quantity 5 g
Specific Gravity 1
Synonym benzoyl chloride, 4-1,1-dimethylethyl, 4-tert-butylbenzoylchloride, p-t-butylbenzoyl chloride, p-tert-butylbenzoyl chloride, benzoyl chloride,4-1,1-dimethylethyl, 4-tert-butyl-benzoyl chloride, 4-tert-butyl benzoyl chloride, 4-t-butylbenzoyl chloride, acmc-1brh5, 4-tertbutylbenzoyl chloride
Solubility Information Solubility in water: readily hydrolysed
Formula Weight 196.68
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 4-tert-Butylbenzoyl chloride, 98%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Harmful if inhaled.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Causes severe skin burns and eye damage.
Contact with water liberates toxic gas.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
Store in a dry place. Store in a closed container.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 216-973-1
TSCA TSCA

RUO – Research Use Only