missing translation for 'onlineSavingsMsg'
Learn More

4-tert-Butylbenzoyl chloride, 98%

Catalog No. AAA1437314
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1437314 25 g
AAA1437322 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1437314 Supplier Thermo Scientific Chemicals Supplier No. A1437314

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1710-98-1 | C11H13ClO | 196.67 g/mol

4-tert-Butylbenzoyl chloride is used in the synthesis of 9,9?-spirobifluorene-bridged bipolar systems containing 1,3,4-oxadiazole-conjugated oligoaryl and triaryllamine moieties. It is also used in the preparation of 2-(4-tert-butylphenyl)-5-(4-methoxyphenyl)-1,3,4-oxadiazole.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-tert-Butylbenzoyl chloride is used in the synthesis of 9,9′-spirobifluorene-bridged bipolar systems containing 1,3,4-oxadiazole-conjugated oligoaryl and triaryllamine moieties. It is also used in the preparation of 2-(4-tert-butylphenyl)-5-(4-methoxyphenyl)-1,3,4-oxadiazole.

Solubility
Reacts with water.

Notes
Moisture sensitive. Store away from oxidizing agents, bases and water/moisture. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas. Keep ignition sources away.

Chemical Identifiers

CAS 1710-98-1
Molecular Formula C11H13ClO
Molecular Weight (g/mol) 196.67
MDL Number MFCD00000695
InChI Key WNLMYNASWOULQY-UHFFFAOYSA-N
PubChem CID 74372
IUPAC Name 4-tert-butylbenzoyl chloride
SMILES CC(C)(C)C1=CC=C(C=C1)C(Cl)=O

Specifications

Density 1.08
Boiling Point 125°C to 126°C (10 mmHg)
Flash Point 87°C (188°F)
Refractive Index 1.5351
Quantity 25 g
UN Number UN3265
Beilstein 775793
Sensitivity Moisture sensitive
Synonym benzoyl chloride, 4-1,1-dimethylethyl, 4-tert-butylbenzoylchloride, p-t-butylbenzoyl chloride, p-tert-butylbenzoyl chloride, benzoyl chloride,4-1,1-dimethylethyl, 4-tert-butyl-benzoyl chloride, 4-tert-butyl benzoyl chloride, 4-t-butylbenzoyl chloride, acmc-1brh5, 4-tertbutylbenzoyl chloride
Solubility Information Reacts with water.
Formula Weight 196.68
Percent Purity 98%
Chemical Name or Material 4-tert-Butylbenzoyl chloride
Show More Show Less
Hazard Category H227-H302+H312+H332-H314-H335
Hazard Statement GHS H Statement
H314-H318-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.
Precautionary Statement P210-P235-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber 216-973-1
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only