missing translation for 'onlineSavingsMsg'
Learn More

4-tert-Butylbenzhydrazide, 98+%

Catalog No. p-7021049
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1322414 25 g
AAA1322422 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1322414 Supplier Thermo Scientific Chemicals Supplier No. A1322414
Only null left

CAS: 43100-38-5 | C11H16N2O | 192.262 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 43100-38-5
Molecular Formula C11H16N2O
Molecular Weight (g/mol) 192.262
MDL Number MFCD00014763
InChI Key XYUFQWDLRLHUPB-UHFFFAOYSA-N
Synonym 4-tert-butylbenzhydrazide, 4-tert-butyl benzohydrazide, benzoic acid, 4-1,1-dimethylethyl-, hydrazide, p-tert-butylbenzohydrazide, 4-tert-butylbenzhyrazide, 4-tert-butylbenzoic acid hydrazide, 4-tert-butylbenzoic hydrazide, 4-tert-butyl benzenecarbohydrazide, 4-tert-butyl benzene-1-carbohydrazide, 4-t-butylbenzhydrazide
PubChem CID 123513
IUPAC Name 4-tert-butylbenzohydrazide
SMILES CC(C)(C)C1=CC=C(C=C1)C(=O)NN

Specifications

Melting Point 127°C to 131°C
Quantity 25 g
Beilstein 1104663
Formula Weight 192.26
Percent Purity ≥98%
Chemical Name or Material 4-tert-Butylbenzhydrazide
Hazard Category Warning
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
EINECSNumber 256-090-9
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only