missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-tert-Butylbenzenesulfonyl Chloride 98.0+%, TCI America™
Supplier: TCI America B198325G
Specifications
| 4-tert-Butylbenzenesulfonyl Chloride | |
| 81°C | |
| C10H13ClO2S | |
| 25 g | |
| 4-t-butylbenzenesulfonyl chloride, 4-tert-butyl benzene-1-sulfonyl chloride, 4-tert-butylbenzene-1-sulfonyl chloride, 4-tert-butylbenzenesulfonylchloride, 4-tert-butyl benzenesulfonyl chloride, 4-tert-butyl-benzenesulfonyl chloride, 4-tert-butylphenylsulfonyl chloride, p-tert-butylbenzenesulfonyl chloride, 4-tert butylbenzenesulfonyl chloride, 4-tert-butyl phenylsulfonyl chloride | |
| CC(C)(C)C1=CC=C(C=C1)S(=O)(=O)Cl | |
| 232.722 | |
| 232.72 | |
| Crystalline Powder |
| 15084-51-2 | |
| 165°C | |
| MFCD00007449 | |
| 3261 | |
| YEZADZMMVHWFIY-UHFFFAOYSA-N | |
| 4-tert-butylbenzenesulfonyl chloride | |
| 139882 | |
| ≥98.0% (T) |
Chemical Identifiers
| 15084-51-2 | |
| 232.722 | |
| YEZADZMMVHWFIY-UHFFFAOYSA-N | |
| 139882 | |
| CC(C)(C)C1=CC=C(C=C1)S(=O)(=O)Cl |
| C10H13ClO2S | |
| MFCD00007449 | |
| 4-t-butylbenzenesulfonyl chloride, 4-tert-butyl benzene-1-sulfonyl chloride, 4-tert-butylbenzene-1-sulfonyl chloride, 4-tert-butylbenzenesulfonylchloride, 4-tert-butyl benzenesulfonyl chloride, 4-tert-butyl-benzenesulfonyl chloride, 4-tert-butylphenylsulfonyl chloride, p-tert-butylbenzenesulfonyl chloride, 4-tert butylbenzenesulfonyl chloride, 4-tert-butyl phenylsulfonyl chloride | |
| 4-tert-butylbenzenesulfonyl chloride |
Safety and Handling
TSCA : No