missing translation for 'onlineSavingsMsg'
Learn More

4-tert-Butylbenzenesulfonyl Chloride 98.0+%, TCI America™

Catalog No. B19835G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B19835G 5 g
B198325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B19835G Supplier TCI America Supplier No. B19835G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 15084-51-2
Molecular Formula C10H13ClO2S
Molecular Weight (g/mol) 232.722
MDL Number MFCD00007449
InChI Key YEZADZMMVHWFIY-UHFFFAOYSA-N
Synonym 4-t-butylbenzenesulfonyl chloride, 4-tert-butyl benzene-1-sulfonyl chloride, 4-tert-butylbenzene-1-sulfonyl chloride, 4-tert-butylbenzenesulfonylchloride, 4-tert-butyl benzenesulfonyl chloride, 4-tert-butyl-benzenesulfonyl chloride, 4-tert-butylphenylsulfonyl chloride, p-tert-butylbenzenesulfonyl chloride, 4-tert butylbenzenesulfonyl chloride, 4-tert-butyl phenylsulfonyl chloride
PubChem CID 139882
IUPAC Name 4-tert-butylbenzenesulfonyl chloride
SMILES CC(C)(C)C1=CC=C(C=C1)S(=O)(=O)Cl

Specifications

Melting Point 81°C
Boiling Point 165°C
Quantity 5 g
UN Number 3261
Formula Weight 232.72
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 4-tert-Butylbenzenesulfonyl Chloride