missing translation for 'onlineSavingsMsg'
Learn More

4-tert-Butylbenzenesulfonyl chloride, 98%

Catalog No. p-7026260
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2217409 10 g
AAB2217418 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2217409 Supplier Thermo Scientific Chemicals Supplier No. B2217409
Only null left

CAS: 15084-51-2 | C10H13ClO2S | 232.722 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 15084-51-2
Molecular Formula C10H13ClO2S
Molecular Weight (g/mol) 232.722
MDL Number MFCD00007449
InChI Key YEZADZMMVHWFIY-UHFFFAOYSA-N
Synonym 4-t-butylbenzenesulfonyl chloride, 4-tert-butyl benzene-1-sulfonyl chloride, 4-tert-butylbenzene-1-sulfonyl chloride, 4-tert-butylbenzenesulfonylchloride, 4-tert-butyl benzenesulfonyl chloride, 4-tert-butyl-benzenesulfonyl chloride, 4-tert-butylphenylsulfonyl chloride, p-tert-butylbenzenesulfonyl chloride, 4-tert butylbenzenesulfonyl chloride, 4-tert-butyl phenylsulfonyl chloride
PubChem CID 139882
IUPAC Name 4-tert-butylbenzenesulfonyl chloride
SMILES CC(C)(C)C1=CC=C(C=C1)S(=O)(=O)Cl

Specifications

Melting Point 80°C to 83°C
Odor Irritating
Quantity 10 g
UN Number UN3261
Beilstein 1104346
Sensitivity Moisture sensitive
Formula Weight 232.73
Percent Purity 98%
Chemical Name or Material 4-tert-Butylbenzenesulfonyl chloride
Hazard Category H314
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only