missing translation for 'onlineSavingsMsg'
Learn More

4-tert-Butylbenzamide, 98%

Catalog No. p-7022548
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1538009 10 g
AAA1538018 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1538009 Supplier Thermo Scientific Chemicals Supplier No. A1538009

CAS: 56108-12-4 | C11H15NO

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 56108-12-4
Molecular Formula C11H15NO
MDL Number MFCD00017139
Synonym 4-tert-butyl benzamide, benzamide, 4-1,1-dimethylethyl, p-tert-butyl benzamide, 4-tert-butyl benzenecarboxamide, unii-5e7907z6bm, 4-1,1-dimethylethyl benzamide, p-t-butylbenzamide, 4-t-butylbenzamide, acmc-20amap, butylbenzenecarboxamide

Specifications

Melting Point 171°C to 175°C
Quantity 10 g
Beilstein 2086692
Formula Weight 177.25
Percent Purity 98%
Chemical Name or Material 4-tert-Butylbenzamide
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only