missing translation for 'onlineSavingsMsg'
Learn More

4'-tert-Butylacetophenone, 98%

Catalog No. p-7021328
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1361503 1 g
AAA1361506 5 g
AAA1361514 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1361503 Supplier Thermo Scientific Chemicals Supplier No. A1361503

CAS: 943-27-1 | C12H16O | 176.259 g/mol

4'-tert-Butylacetophenone is used in the synthesis of 2-pyridone derivatives.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4′-tert-Butylacetophenone is used in the synthesis of 2-pyridone derivatives.

Solubility
Soluble in alcohol. Insoluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 943-27-1
Molecular Formula C12H16O
Molecular Weight (g/mol) 176.259
MDL Number MFCD00017256
InChI Key UYFJYGWNYQCHOB-UHFFFAOYSA-N
Synonym 4'-tert-butylacetophenone, 1-4-tert-butylphenyl ethanone, 4-tert-butylacetophenone, 1-4-tert-butyl phenyl ethanone, p-tert-butylacetophenone, ethanone, 1-4-1,1-dimethylethyl phenyl, acetophenone, 4'-tert-butyl, 1-4-tert-butyl phenyl ethan-1-one, p-t-butylacetophenone, 1-4-tert-butyl-phenyl-ethanone
PubChem CID 13669
IUPAC Name 1-(4-tert-butylphenyl)ethanone
SMILES CC(=O)C1=CC=C(C=C1)C(C)(C)C

Specifications

Melting Point 17°C to 18°C (dimorphic)
Density 0.964
Boiling Point 107°C to 108°C (5 mmHg)
Flash Point >100°C (212°F)
Refractive Index 1.52
Quantity 1 g
Solubility Information Soluble in alcohol. Insoluble in water.
Formula Weight 176.26
Percent Purity 98%
Chemical Name or Material 4'-tert-Butylacetophenone
Hazard Category H226-H315-H319-H335
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
DOTInformation Hazard Class: 3; Packaging Group: III
EINECSNumber 213-399-3
TSCA Yes

RUO – Research Use Only