missing translation for 'onlineSavingsMsg'
Learn More

4'-tert-Butylacetophenone, 98%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 308740010

 View more versions of this product

Catalog No. AC308740010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

4'-tert-Butylacetophenone
943-27-1
100.0
107.0°C to 108.0°C (5.0 mmHg)
Authentic
Glass bottle
1.5190 to 1.5220
MFCD00017256
0.96
Solubility in water: insoluble. Other solubilities: soluble in alkanes,methylene chloride,ethanol,,2-propanol,acetone
CC(=O)C1=CC=C(C=C1)C(C)(C)C
176.26
176.26
98%
97.5
0.9600g/mL
29°C
97.5% min. (GC)
C12H16O
(CH3)3CC6H4COCH3
1g
4'-tert-butylacetophenone, 1-4-tert-butylphenyl ethanone, 4-tert-butylacetophenone, 1-4-tert-butyl phenyl ethanone, p-tert-butylacetophenone, ethanone, 1-4-1,1-dimethylethyl phenyl, acetophenone, 4'-tert-butyl, 1-4-tert-butyl phenyl ethan-1-one, p-t-butylacetophenone, 1-4-tert-butyl-phenyl-ethanone
UYFJYGWNYQCHOB-UHFFFAOYSA-N
1-(4-tert-butylphenyl)ethanone
13669
98%

Chemical Identifiers

943-27-1
176.26
UYFJYGWNYQCHOB-UHFFFAOYSA-N
13669
CC(=O)C1=CC=C(C=C1)C(C)(C)C
C12H16O
MFCD00017256
4'-tert-butylacetophenone, 1-4-tert-butylphenyl ethanone, 4-tert-butylacetophenone, 1-4-tert-butyl phenyl ethanone, p-tert-butylacetophenone, ethanone, 1-4-1,1-dimethylethyl phenyl, acetophenone, 4'-tert-butyl, 1-4-tert-butyl phenyl ethan-1-one, p-t-butylacetophenone, 1-4-tert-butyl-phenyl-ethanone
1-(4-tert-butylphenyl)ethanone
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Wash face,hands and any exposed skin thoroughly

GHS Signal Word: Warning

EINECSNumber : 213-399-3

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only