missing translation for 'onlineSavingsMsg'
Learn More

4-tert-Butyl N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-D-aspartate 98.0+%, TCI America™

Catalog No. B46191G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B46191G 1 g
B46195G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B46191G Supplier TCI America Supplier No. B46191G

Chemical Identifiers

CAS 112883-39-3
Molecular Formula C23H25NO6
Molecular Weight (g/mol) 411.45
MDL Number MFCD00077050
InChI Key FODJWPHPWBKDON-UHFFFAOYNA-N
Synonym fmoc-d-asp otbu-oh, r-2-9h-fluoren-9-yl methoxy carbonyl amino-4-tert-butoxy-4-oxobutanoic acid, fmoc-d-asp obut-oh, fmoc-d-aspartic acid beta-tert-butyl ester, n-alpha-fmoc-d-aspartic acid beta-t-butyl ester, 2r-4-tert-butoxy-2-9h-fluoren-9-ylmethoxy carbonyl amino-4-oxobutanoic acid, fmoc-dasp otbu, ambotzfaa1309, fmoc-dasp otbu-oh, pubchem10009
PubChem CID 7018816
IUPAC Name 4-(tert-butoxy)-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-4-oxobutanoic acid
SMILES CC(C)(C)OC(=O)CC(NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O

Specifications

Melting Point 149°C
Color White
Quantity 1 g
Formula Weight 411.45
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-tert-Butyl N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-D-aspartate