missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-tert-Butyl L-Aspartate 98.0+%, TCI America™
$102.03 - $392.74
Chemical Identifiers
| CAS | 3057-74-7 |
|---|---|
| Molecular Formula | C8H15NO4 |
| Molecular Weight (g/mol) | 189.211 |
| MDL Number | MFCD00038577 |
| InChI Key | MXWMFBYWXMXRPD-YFKPBYRVSA-N |
| Synonym | h-asp otbu-oh, l-aspartic acid 4-tert-butyl ester, h-asp obut-oh, 4-tert-butyl hydrogen l-aspartate, 2s-2-amino-4-tert-butoxy-4-oxobutanoic acid, s-2-amino-4-tert-butoxy-4-oxobutanoic acid, aspartic acid, 4-tert-butyl ester, asp but, asp obut, asp otbu oh |
| PubChem CID | 7010570 |
| IUPAC Name | (2S)-2-amino-4-[(2-methylpropan-2-yl)oxy]-4-oxobutanoic acid |
| SMILES | CC(C)(C)OC(=O)CC(C(=O)O)N |
Chemical Identifiers
| 3057-74-7 | |
| 189.211 | |
| MXWMFBYWXMXRPD-YFKPBYRVSA-N | |
| 7010570 | |
| CC(C)(C)OC(=O)CC(C(=O)O)N |
| C8H15NO4 | |
| MFCD00038577 | |
| h-asp otbu-oh, l-aspartic acid 4-tert-butyl ester, h-asp obut-oh, 4-tert-butyl hydrogen l-aspartate, 2s-2-amino-4-tert-butoxy-4-oxobutanoic acid, s-2-amino-4-tert-butoxy-4-oxobutanoic acid, aspartic acid, 4-tert-butyl ester, asp but, asp obut, asp otbu oh | |
| (2S)-2-amino-4-[(2-methylpropan-2-yl)oxy]-4-oxobutanoic acid |
Specifications
| 3057-74-7 | |
| White | |
| MFCD00038577 | |
| h-asp otbu-oh, l-aspartic acid 4-tert-butyl ester, h-asp obut-oh, 4-tert-butyl hydrogen l-aspartate, 2s-2-amino-4-tert-butoxy-4-oxobutanoic acid, s-2-amino-4-tert-butoxy-4-oxobutanoic acid, aspartic acid, 4-tert-butyl ester, asp but, asp obut, asp otbu oh | |
| CC(C)(C)OC(=O)CC(C(=O)O)N | |
| 189.211 | |
| 189.21 | |
| Crystalline Powder |
| 189°C | |
| C8H15NO4 | |
| 1 g | |
| MXWMFBYWXMXRPD-YFKPBYRVSA-N | |
| (2S)-2-amino-4-[(2-methylpropan-2-yl)oxy]-4-oxobutanoic acid | |
| 7010570 | |
| ≥98.0% (T) | |
| 4-tert-Butyl L-Aspartate |
Safety and Handling
TSCA : No