missing translation for 'onlineSavingsMsg'
Learn More

4'-tert-Butyl-4-chlorobutyrophenone 95.0+%, TCI America™

Catalog No. B314425G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B314425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B314425G Supplier TCI America Supplier No. B314425G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 43076-61-5
Molecular Formula C14H19ClO
Molecular Weight (g/mol) 238.755
MDL Number MFCD00018996
InChI Key RLKSQLJFGCDUOX-UHFFFAOYSA-N
Synonym 4'-tert-butyl-4-chlorobutyrophenone, 1-4-tert-butyl phenyl-4-chlorobutan-1-one, 4'-tertbutyl-4-chlorobutyrophenone, 1-4-tert-butylphenyl-4-chlorobutan-1-one, acmc-209up0, dsstox_cid_31534, dsstox_rid_97419, dsstox_gsid_57745, 4-chloro-1-4-1,1-dimethylethyl phenyl-1-butanone
PubChem CID 2723694
IUPAC Name 1-(4-tert-butylphenyl)-4-chlorobutan-1-one
SMILES CC(C)(C)C1=CC=C(C=C1)C(=O)CCCCl

Specifications

Melting Point 46°C
Color Yellow
Boiling Point 155°C
Quantity 25 g
Formula Weight 238.76
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4′-tert-Butyl-4-chlorobutyrophenone