missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-tert-Butyl-1-(ethoxycarbonylmethoxy)thiacalix[4]arene 94.0+%, TCI America™
$160.30 - $477.19
Chemical Identifiers
| CAS | 210706-03-9 |
|---|---|
| Molecular Formula | C56H72O12S4 |
| Molecular Weight (g/mol) | 1065.42 |
| MDL Number | MFCD06797059 |
| InChI Key | IGVIIKFHGZNQFL-UHFFFAOYSA-N |
| Synonym | Tetra-tert-butyl[tetrakis(ethoxycarbonylmethoxy)]tetrathiacalix[4]arene |
| PubChem CID | 5248766 |
| IUPAC Name | ethyl 2-{[5,11,17,23-tetra-tert-butyl-26,27,28-tris(2-ethoxy-2-oxoethoxy)-2,8,14,20-tetrathiapentacyclo[19.3.1.1³,â·.1â¹,¹³.1¹âµ,¹â¹]octacosa-1(25),3(28),4,6,9(27),10,12,15,17,19(26),21,23-dodecaen-25-yl]oxy}acetate |
| SMILES | CCOC(=O)COC1=C2SC3=CC(=CC(SC4=C(OCC(=O)OCC)C(SC5=C(OCC(=O)OCC)C(SC1=CC(=C2)C(C)(C)C)=CC(=C5)C(C)(C)C)=CC(=C4)C(C)(C)C)=C3OCC(=O)OCC)C(C)(C)C |
Chemical Identifiers
| 210706-03-9 | |
| 1065.42 | |
| IGVIIKFHGZNQFL-UHFFFAOYSA-N | |
| 5248766 | |
| CCOC(=O)COC1=C2SC3=CC(=CC(SC4=C(OCC(=O)OCC)C(SC5=C(OCC(=O)OCC)C(SC1=CC(=C2)C(C)(C)C)=CC(=C5)C(C)(C)C)=CC(=C4)C(C)(C)C)=C3OCC(=O)OCC)C(C)(C)C |
| C56H72O12S4 | |
| MFCD06797059 | |
| Tetra-tert-butyl[tetrakis(ethoxycarbonylmethoxy)]tetrathiacalix[4]arene | |
| ethyl 2-{[5,11,17,23-tetra-tert-butyl-26,27,28-tris(2-ethoxy-2-oxoethoxy)-2,8,14,20-tetrathiapentacyclo[19.3.1.1³,â·.1â¹,¹³.1¹âµ,¹â¹]octacosa-1(25),3(28),4,6,9(27),10,12,15,17,19(26),21,23-dodecaen-25-yl]oxy}acetate |
Specifications
| 210706-03-9 | |
| C56H72O12S4 | |
| 1 g | |
| IGVIIKFHGZNQFL-UHFFFAOYSA-N | |
| ethyl 2-{[5,11,17,23-tetra-tert-butyl-26,27,28-tris(2-ethoxy-2-oxoethoxy)-2,8,14,20-tetrathiapentacyclo[19.3.1.1³,â·.1â¹,¹³.1¹âµ,¹â¹]octacosa-1(25),3(28),4,6,9(27),10,12,15,17,19(26),21,23-dodecaen-25-yl]oxy}acetate | |
| 5248766 | |
| ≥94.0% (HPLC) | |
| 4-tert-Butyl-1-(ethoxycarbonylmethoxy)thiacalix[4]arene |
| White-Yellow | |
| MFCD06797059 | |
| Tetra-tert-butyl[tetrakis(ethoxycarbonylmethoxy)]tetrathiacalix[4]arene | |
| CCOC(=O)COC1=C2SC3=CC(=CC(SC4=C(OCC(=O)OCC)C(SC5=C(OCC(=O)OCC)C(SC1=CC(=C2)C(C)(C)C)=CC(=C5)C(C)(C)C)=CC(=C4)C(C)(C)C)=C3OCC(=O)OCC)C(C)(C)C | |
| 1065.42 | |
| 1065.42 | |
| Crystalline Powder |
Safety and Handling
TSCA : No