missing translation for 'onlineSavingsMsg'
Learn More

4-(tert-Butyl)-1,3-thiazol-2-amine, 97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals CD08308EB

Quantity 25g

 View more versions of this product

Catalog No. CD08308EB

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Chemical Identifiers

74370-93-7
156.247
CUWZBHVYLVGOAB-UHFFFAOYSA-N
2734202
CC(C)(C)C1=CSC(=N1)N
C7H12N2S
MFCD00051644
2-amino-4-tert-butylthiazole, 4-tert-butylthiazol-2-ylamine, 4-tert-butyl thiazol-2-amine, 4-tert-butyl-thiazol-2-ylamine, 2-amino-4-t-butylthiazole, 4-tert-butyl-1,3-thiazol-2-amine, 4-tert-butylthiazol-2-amine, 4-tert-butyl-1,3-thiazol-2-ylamine, 2-thiazolamine, 4-1,1-dimethylethyl, 4-tert-butyl-1,3-thiazole-2-ylamine
4-tert-butyl-1,3-thiazol-2-amine

Specifications

4-(tert-Butyl)-1, 3-thiazol-2-amine
96.5
97%
25g
2-amino-4-tert-butylthiazole, 4-tert-butylthiazol-2-ylamine, 4-tert-butyl thiazol-2-amine, 4-tert-butyl-thiazol-2-ylamine, 2-amino-4-t-butylthiazole, 4-tert-butyl-1,3-thiazol-2-amine, 4-tert-butylthiazol-2-amine, 4-tert-butyl-1,3-thiazol-2-ylamine, 2-thiazolamine, 4-1,1-dimethylethyl, 4-tert-butyl-1,3-thiazole-2-ylamine
CC(C)(C)C1=CSC(=N1)N
156.247
156.25
74370-93-7
100.0
C7H12N2S
MFCD00051644
CUWZBHVYLVGOAB-UHFFFAOYSA-N
4-tert-butyl-1,3-thiazol-2-amine
2734202
97%
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Harmful if swallowed.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS
Documents

Documents