missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-(tert-Butoxycarbonylamino)piperidine 98.0+%, TCI America™
$36.57 - $51.98
Chemical Identifiers
| CAS | 73874-95-0 |
|---|---|
| Molecular Formula | C10H21N2O2 |
| Molecular Weight (g/mol) | 201.29 |
| MDL Number | MFCD00798171 |
| InChI Key | CKXZPVPIDOJLLM-UHFFFAOYSA-O |
| Synonym | 4-boc-aminopiperidine, tert-butyl piperidin-4-ylcarbamate, 4-n-boc-aminopiperidine, 4-boc-amino piperidine, 4-n-boc-amino piperidine, tert-butyl n-piperidin-4-yl carbamate, 4-tert-butoxycarbonylamino piperidine, 4-n-boc-amino-piperidine, 4-boc-amino-piperidine, 4-n-boc amino piperidine |
| PubChem CID | 723833 |
| IUPAC Name | 4-{[(tert-butoxy)carbonyl]amino}piperidin-1-ium |
| SMILES | CC(C)(C)OC(=O)NC1CC[NH2+]CC1 |
Chemical Identifiers
| 73874-95-0 | |
| 201.29 | |
| CKXZPVPIDOJLLM-UHFFFAOYSA-O | |
| 723833 | |
| CC(C)(C)OC(=O)NC1CC[NH2+]CC1 |
| C10H21N2O2 | |
| MFCD00798171 | |
| 4-boc-aminopiperidine, tert-butyl piperidin-4-ylcarbamate, 4-n-boc-aminopiperidine, 4-boc-amino piperidine, 4-n-boc-amino piperidine, tert-butyl n-piperidin-4-yl carbamate, 4-tert-butoxycarbonylamino piperidine, 4-n-boc-amino-piperidine, 4-boc-amino-piperidine, 4-n-boc amino piperidine | |
| 4-{[(tert-butoxy)carbonyl]amino}piperidin-1-ium |
Specifications
| 159°C | |
| C10H21N2O2 | |
| 4-boc-aminopiperidine, tert-butyl piperidin-4-ylcarbamate, 4-n-boc-aminopiperidine, 4-boc-amino piperidine, 4-n-boc-amino piperidine, tert-butyl n-piperidin-4-yl carbamate, 4-tert-butoxycarbonylamino piperidine, 4-n-boc-amino-piperidine, 4-boc-amino-piperidine, 4-n-boc amino piperidine | |
| CC(C)(C)OC(=O)NC1CC[NH2+]CC1 | |
| 1 g | |
| 723833 | |
| ≥98.0% (GC,T) | |
| 4-(tert-Butoxycarbonylamino)piperidine |
| 73874-95-0 | |
| MFCD00798171 | |
| CKXZPVPIDOJLLM-UHFFFAOYSA-O | |
| 4-{[(tert-butoxy)carbonyl]amino}piperidin-1-ium | |
| 201.29 | |
| 200.28 | |
| Crystalline Powder |
Safety and Handling
TSCA : No