missing translation for 'onlineSavingsMsg'
Learn More

4-(tert-Butoxycarbonylamino)piperidine 98.0+%, TCI America™

Catalog No. B23195G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B23195G 5 g
B23191G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B23195G Supplier TCI America Supplier No. B23195G

Chemical Identifiers

CAS 73874-95-0
Molecular Formula C10H21N2O2
Molecular Weight (g/mol) 201.29
MDL Number MFCD00798171
InChI Key CKXZPVPIDOJLLM-UHFFFAOYSA-O
Synonym 4-boc-aminopiperidine, tert-butyl piperidin-4-ylcarbamate, 4-n-boc-aminopiperidine, 4-boc-amino piperidine, 4-n-boc-amino piperidine, tert-butyl n-piperidin-4-yl carbamate, 4-tert-butoxycarbonylamino piperidine, 4-n-boc-amino-piperidine, 4-boc-amino-piperidine, 4-n-boc amino piperidine
PubChem CID 723833
IUPAC Name 4-{[(tert-butoxy)carbonyl]amino}piperidin-1-ium
SMILES CC(C)(C)OC(=O)NC1CC[NH2+]CC1

Specifications

Melting Point 159°C
Quantity 5 g
Formula Weight 200.28
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-(tert-Butoxycarbonylamino)piperidine