Learn More
4-[(tert-Butoxycarbonylamino)methyl]phenylboronic acid pinacol ester, 97%
CAS: 330794-35-9 | C18H28BNO4 | 333.24 g/mol
$225.29 - $225.29
Chemical Identifiers
| CAS | 330794-35-9 |
|---|---|
| Molecular Formula | C18H28BNO4 |
| Molecular Weight (g/mol) | 333.24 |
| MDL Number | MFCD06409942 |
| InChI Key | CUDCEJRRWNIPDL-UHFFFAOYSA-N |
| Synonym | 4-n-boc-aminomethyl phenylboronic acid pinacol ester, tert-butyl 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzylcarbamate, 4-n-boc-amino methyl phenylboronic acid pinacol ester, 4-tert-butoxycarbonylaminomethyl phenylboronic acid, pinacol ester, tert-butyl n-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl methyl carbamate, 4-tert-butoxycarbonylaminomethyl phenylboronic acid pinacol ester, 4-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-benzyl-carbamic acid tert-butyl ester, pubchem5247, 4-n-boc-aminomethyl phenylboronic acid,pinacol ester |
| PubChem CID | 3415442 |
| IUPAC Name | tert-butyl N-[[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]methyl]carbamate |
| SMILES | CC(C)(C)OC(=O)NCC1=CC=C(C=C1)B1OC(C)(C)C(C)(C)O1 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AC444640010
|
Thermo Scientific Chemicals
444640010 |
1 g |
Each for $225.29
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 330794-35-9 | |
| 333.24 | |
| CUDCEJRRWNIPDL-UHFFFAOYSA-N | |
| 3415442 | |
| CC(C)(C)OC(=O)NCC1=CC=C(C=C1)B1OC(C)(C)C(C)(C)O1 |
| C18H28BNO4 | |
| MFCD06409942 | |
| 4-n-boc-aminomethyl phenylboronic acid pinacol ester, tert-butyl 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzylcarbamate, 4-n-boc-amino methyl phenylboronic acid pinacol ester, 4-tert-butoxycarbonylaminomethyl phenylboronic acid, pinacol ester, tert-butyl n-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl methyl carbamate, 4-tert-butoxycarbonylaminomethyl phenylboronic acid pinacol ester, 4-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-benzyl-carbamic acid tert-butyl ester, pubchem5247, 4-n-boc-aminomethyl phenylboronic acid,pinacol ester | |
| tert-butyl N-[[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]methyl]carbamate |
Specifications
| 330794-35-9 | |
| 96% min. (GC) | |
| C18H28BNO4 | |
| 1 g | |
| CUDCEJRRWNIPDL-UHFFFAOYSA-N | |
| tert-butyl N-[[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]methyl]carbamate | |
| 3415442 | |
| 97% |
| 68.0°C to 69.0°C | |
| Glass Bottle | |
| MFCD06409942 | |
| 4-n-boc-aminomethyl phenylboronic acid pinacol ester, tert-butyl 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzylcarbamate, 4-n-boc-amino methyl phenylboronic acid pinacol ester, 4-tert-butoxycarbonylaminomethyl phenylboronic acid, pinacol ester, tert-butyl n-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl methyl carbamate, 4-tert-butoxycarbonylaminomethyl phenylboronic acid pinacol ester, 4-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-benzyl-carbamic acid tert-butyl ester, pubchem5247, 4-n-boc-aminomethyl phenylboronic acid,pinacol ester | |
| CC(C)(C)OC(=O)NCC1=CC=C(C=C1)B1OC(C)(C)C(C)(C)O1 | |
| 333.24 | |
| 333.24 | |
| 4-[(tert-Butoxycarbonylamino)methyl]phenylboronic acid pinacol ester |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF
GHS Signal Word: Warning
RUO – Research Use Only