missing translation for 'onlineSavingsMsg'
Learn More

4-[(tert-Butoxycarbonylamino)methyl]phenylboronic acid pinacol ester, 97%

Catalog No. AC444640010
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC444640010 1 g
AC444640050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC444640010 Supplier Thermo Scientific Chemicals Supplier No. 444640010
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 330794-35-9 | C18H28BNO4 | 333.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 330794-35-9
Molecular Formula C18H28BNO4
Molecular Weight (g/mol) 333.24
MDL Number MFCD06409942
InChI Key CUDCEJRRWNIPDL-UHFFFAOYSA-N
Synonym 4-n-boc-aminomethyl phenylboronic acid pinacol ester, tert-butyl 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzylcarbamate, 4-n-boc-amino methyl phenylboronic acid pinacol ester, 4-tert-butoxycarbonylaminomethyl phenylboronic acid, pinacol ester, tert-butyl n-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl methyl carbamate, 4-tert-butoxycarbonylaminomethyl phenylboronic acid pinacol ester, 4-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-benzyl-carbamic acid tert-butyl ester, pubchem5247, 4-n-boc-aminomethyl phenylboronic acid,pinacol ester
PubChem CID 3415442
IUPAC Name tert-butyl N-[[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]methyl]carbamate
SMILES CC(C)(C)OC(=O)NCC1=CC=C(C=C1)B1OC(C)(C)C(C)(C)O1

Specifications

Melting Point 68.0°C to 69.0°C
Assay Percent Range 96% min. (GC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 333.24
Percent Purity 97%
Chemical Name or Material 4-[(tert-Butoxycarbonylamino)methyl]phenylboronic acid pinacol ester
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF

RUO – Research Use Only