missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Sulfothiacalix[4]arene Sodium Salt 98.0+%, TCI America™
Supplier: TCI America S04775G
Specifications
| 4-Sulfothiacalix[4]arene Sodium Salt | |
| 390°C | |
| C24H16O16S8 | |
| 5 g | |
| 904.78 | |
| Crystalline Powder |
| 211561-04-5 | |
| White | |
| MFCD06797166 | |
| Tetrasodium Tetrathiacalix[4]arenetetrasulfonate, Thiacalix[4]arenetetrasulfonic Acid Tetrasodium Salt | |
| ≥98.0% (HPLC) |
Chemical Identifiers
| 211561-04-5 | |
| C24H16O16S8 | |
| MFCD06797166 | |
| Tetrasodium Tetrathiacalix[4]arenetetrasulfonate, Thiacalix[4]arenetetrasulfonic Acid Tetrasodium Salt |
Safety and Handling
TSCA : No