missing translation for 'onlineSavingsMsg'
Learn More

4-Sulfocalix[6]arene Hydrate 95.0+%, TCI America™

Catalog No. S04701G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S04701G 1 g
S04705G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S04701G Supplier TCI America Supplier No. S04701G
Only null left

Chemical Identifiers

CAS 102088-39-1
Molecular Formula C42H36O24S6
Molecular Weight (g/mol) 1117.09
MDL Number MFCD00216907,MFCD00143376
InChI Key BXOUPTJVBGEDIR-UHFFFAOYSA-N
Synonym 4-sulfocalix 6 arene, 4-sulfocalix 6 arene hydrate, hexahydroxycalix 6 arenehexasulfonic acid, 4-sulfonic calix 6 arene, acmc-20m53e, calix 6 arene-p-sulfonate, hexasulfo hexahydroxy calix 6 arene
PubChem CID 2724887
IUPAC Name 37,38,39,40,41,42-hexahydroxyheptacyclo[31.3.1.1³,⁷.1⁹,¹³.1¹⁵,¹⁹.1²¹,²⁵.1²⁷,³¹]dotetraconta-1(37),3,5,7(42),9,11,13(41),15(40),16,18,21(39),22,24,27(38),28,30,33,35-octadecaene-5,11,17,23,29,35-hexasulfonic acid
SMILES OC1=C2CC3=CC(=CC(CC4=CC(=CC(CC5=C(O)C(CC6=C(O)C(CC7=C(O)C(CC1=CC(=C2)S(O)(=O)=O)=CC(=C7)S(O)(=O)=O)=CC(=C6)S(O)(=O)=O)=CC(=C5)S(O)(=O)=O)=C4O)S(O)(=O)=O)=C3O)S(O)(=O)=O

Specifications

Melting Point 246°C
Quantity 1 g
Formula Weight 1117.09
Percent Purity ≥95.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Sulfocalix[6]arene Hydrate